Skip to Content
Merck
All Photos(1)

Key Documents

8.18374

Sigma-Aldrich

Sodium tert-butylate

for synthesis

Synonym(s):

Sodium tert-butylate, Sodium-t-butoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CONa
CAS Number:
Molecular Weight:
96.10
MDL number:
UNSPSC Code:
12352107
EC Index Number:
212-741-9
NACRES:
NA.22

grade

for synthesis

Quality Level

Assay

≥96.0% (acidimetric)

form

powder

mp

263 °C

bulk density

400 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[O-]C(C)(C)C

InChI

1S/C4H9O.Na/c1-4(2,3)5;/h1-3H3;/q-1;+1

InChI key

MFRIHAYPQRLWNB-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 96.0 %
Free NaOH: ≤ 2.0 %
Identity (IR): passes test

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Self-heat. 1 - Skin Corr. 1B

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service