Skip to Content
Merck
All Photos(3)

Key Documents

8.18056

Sigma-Aldrich

Palladium(II) acetate

47% Pd, for synthesis

Synonym(s):

Palladium(II) acetate (47% Pd), Palladium diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Pd
CAS Number:
Molecular Weight:
224.51
MDL number:
UNSPSC Code:
12352103
EC Index Number:
222-164-4
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

Palladium(II) acetate (47% Pd), for synthesis

Quality Level

form

powder

potency

≥5110 mg/kg LD50, oral (Rat)

reaction suitability

reagent type: oxidant

pH

2-3 (20 °C in H2O, aqueous suspension)

mp

205 °C

storage temp.

2-30°C

SMILES string

[Pd](OC(=O)C)OC(=O)C

InChI

1S/2C2H4O2.Pd/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

YJVFFLUZDVXJQI-UHFFFAOYSA-L

Application

<ul>
<li><strong>Palladium(II) Acetate in Cross-Coupling Reactions:</strong> palladium(II) acetate is used in Suzuki-Miyaura cross-coupling reactions conducted in semi-aqueous media, emphasizing its effectiveness in homogeneous catalysis and potential for diverse organic syntheses (Berm and uacute;dez-Puente et al., 2022).</li>
<li><strong>Microbial Viability Assay Development:</strong> Cechova et al. developed a viability assay that utilizes a palladium compound to treat and detect viable Mycobacterium avium subsp. paratuberculosis cells, illustrating the adaptability of palladium(II) acetate in pharmaceutical research beyond typical catalytic applications (Cechova et al., 2022).</li>
<li><strong>Advanced Catalytic Processes:</strong> palladium(II) acetate is used during the palladium-catalyzed cascade reaction forming 4-alkynyloxazolones from ynamides, showcasing the catalyst and rsquo;s versatility and efficiency in high-precision organic synthesis (Goulart et al., 2022).</li>
</ul>

Analysis Note

Assay (ICP-OES): ≥ 99.0 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service