Skip to Content
Merck
All Photos(1)

Key Documents

8.14506

Sigma-Aldrich

Chlorobenzene

special quality for synthesis

Synonym(s):

Chlorobenzene, Phenyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Cl
CAS Number:
Molecular Weight:
112.56
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-628-5
NACRES:
NA.21
Assay:
≥99.5% (GC)
bp:
132 °C/1013 hPa
vapor pressure:
12 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

12 hPa ( 20 °C)

Quality Level

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

590 °C

potency

2000-4000 mg/kg LD50, oral (Rat)

expl. lim.

1.3-11 % (v/v)

pH

7 (20 °C)

bp

132 °C/1013 hPa

mp

-45 °C

transition temp

flash point 27 °C

density

1.11 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Clc1ccccc1

InChI

1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H

InChI key

MVPPADPHJFYWMZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.5 % (a/a)
Density (d 20 °C/ 4 °C): 1.105 - 1.107
Water (K. F.): ≤ 0.03 %
Free acid: ≤ 0.0002 meq/ml
Free alkali: ≤ 0.0002 meq/ml
Identity (IR): passes test

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service