Skip to Content
Merck

Skip To

8.11684

Crown ether/18-Crown-6

greener alternative

for synthesis

Synonym(s):

Crown ether/18-Crown-6, 1,4,7,10,13,16-Hexaoxacyclooctadecane, 18-Crown-6

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H24O6
CAS Number:
Molecular Weight:
264.32
UNSPSC Code:
12352005
EC Index Number:
241-473-5
NACRES:
NA.22
MDL number:
Assay:
98.0 %, ≥98.0% (GC)
Form:
solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

98.0 %, ≥98.0% (GC)

form

solid

potency

525 mg/kg LD50, oral (Rat), 3888 mg/kg LD50, skin (Rabbit)

reaction suitability

core: crown ether

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

36-39 °C

transition temp

flash point 109 °C

greener alternative category

storage temp.

2-30°C

SMILES string

O1CCOCCOCCOCCOCCOCC1

InChI

1S/C12H24O6/c1-2-14-5-6-16-9-10-18-12-11-17-8-7-15-4-3-13-1/h1-12H2

InChI key

XEZNGIUYQVAUSS-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

  • 18-Crown-6 coordinated metal halides with bright luminescence and nonlinear optical effects: This study explores the luminescence and nonlinear optical properties of crown-ether coordination compounds, indicating potential applications in optoelectronics and photonics (Merzlyakova et al., 2021).
  • 18-Crown-6–sodium cholate complex: thermochemistry, structure, and stability: The study investigates the complex formation between 18-Crown-6 and sodium cholate, offering insights into the thermodynamic stability and structural characteristics of this complex (Mihelj et al., 2014).
  • Alkali Metal Ion Recognition by 18-Crown-6 in Aqueous Solutions: This article presents evidence from local structures on how 18-Crown-6 selectively recognizes and binds alkali metal ions, useful for designing more effective ion-selective sensors (Jing et al., 2023).
  • Influence of 18-Crown-6 Ether Coordination on the Catalytic Activity of Potassium and Calcium Diarylphosphinites: Exploring how 18-Crown-6 ethers influence the catalytic activity in hydrophosphorylation reactions, this research provides a pathway to understanding the role of crown ethers in catalysis (Härling et al., 2017).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 36 °C
Melting range (upper value): ≤ 41 °C
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.118668.117318.13755
assay

98.0 %, ≥98.0% (GC)

assay

≥97.0% (GC), 97.5%

assay

99.9 %, ≥98.0% (GC)

assay

99%

reaction suitability

core: crown ether

reaction suitability

core: crown ether

reaction suitability

core: crown ether

reaction suitability

core: crown ether

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

solid

form

powder

form

powder

form

powder

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

mp

36-39 °C

mp

46-50 °C

mp

158-161 °C

mp

100-103 °C


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters