Skip to Content
Merck
All Photos(1)

Key Documents

8.07839

Sigma-Aldrich

Antimony(III) fluoride

for synthesis

Synonym(s):

Antimony(III) fluoride, Antimony trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
F3Sb
CAS Number:
Molecular Weight:
178.76
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-009-2
NACRES:
NA.22
form:
crystals
solubility:
4430 g/L (Hydrolysis)

Quality Level

form

crystals

bp

376 °C/1013 hPa

mp

292 °C

solubility

4430 g/L (Hydrolysis)

density

4.38 g/cm3 at 20 °C

bulk density

1500 kg/m3

storage temp.

2-30°C

SMILES string

[Sb+3].[F-].[F-].[F-]

InChI

1S/3FH.Sb/h3*1H;/q;;;+3/p-3

InChI key

GUNJVIDCYZYFGV-UHFFFAOYSA-K

Analysis Note

Assay (iodometric): ≥ 99.0 %
Identity (ICP-OES): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service