Skip to Content
Merck
All Photos(1)

Key Documents

8.04012

Sigma-Aldrich

Furfural

for synthesis

Synonym(s):

Furfural, 2-Furancarbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4O2
CAS Number:
Molecular Weight:
96.08
MDL number:
UNSPSC Code:
12352114
EC Index Number:
202-627-7
NACRES:
NA.22
bp:
162 °C/1013 hPa
vapor pressure:
1 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

1 hPa ( 20 °C)

Quality Level

Assay

≥98% (GC)

form

liquid

autoignition temp.

315 °C

potency

65 mg/kg LD50, oral (Rat)

expl. lim.

2.1-19.3 % (v/v)

bp

162 °C/1013 hPa

mp

-37 °C

transition temp

flash point 60 °C

solubility

83 g/L

density

1.16 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[o]1c(ccc1)C=O

InChI

1S/C5H4O2/c6-4-5-2-1-3-7-5/h1-4H

InChI key

HYBBIBNJHNGZAN-UHFFFAOYSA-N

Application

  • Electrocatalytic conversion of furfural: Zhu et al. reviewed the mechanisms, catalysts, and perspectives on the electrocatalytic conversion of biomass-derived furan compounds, including furfural. This process emphasizes the potential of furfural in green chemistry and chemical process optimization (Zhu et al., 2024).
  • Biocatalytic production of furfural: Gong et al. explored the use of electron-rich Ru supported on N-doped coffee biochar for the selective reductive amination of furfural to furfurylamine, highlighting a novel method for furfural utilization in pharmaceutical synthesis and biomass chemical production (Gong et al., 2024).
  • Use of furfural in detoxification: Wang et al. discussed the application of microalgae in the detoxification of lignocellulosic pretreatment wastewater, demonstrating furfural′s role in the sustainable management and resource utilization of wastewater, contributing to environmental and chemical process optimization (Wang et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.158 - 1.160
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service