Skip to Content
Merck
All Photos(1)

Key Documents

8.03235

Sigma-Aldrich

N,N-Dimethylacetamide

for synthesis

Synonym(s):

N,N-Dimethylacetamide, Acetic acid dimethylamide, Acetyldimethylamine, DMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
Molecular Weight:
87.12
MDL number:
UNSPSC Code:
12352111
EC Index Number:
204-826-4
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
165-166 °C/1013 hPa
vapor pressure:
1.76 hPa ( 20 °C)

grade

for synthesis
synthesis grade

Quality Level

vapor pressure

1.76 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

400 °C

potency

5830 mg/kg LD50, oral (Rat)

expl. lim.

1.7-11.5 % (v/v)

dilution

(for synthesis)

pH

4 (20 °C, 200 g/L in H2O)

bp

165-166 °C/1013 hPa

mp

-20 °C

transition temp

flash point 64 °C

solubility

soluble >1000 g/L

density

0.94 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(C)(C)C(=O)C

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylacetamide (DMAc) is a water-miscible organic solvent used in the synthesis of styrene, vinyl resin, linear polyester, acrylic fiber, plastic, pesticide, and synthetic leather. It participates in dehydrogenation, dehydration, halogenation, deprotonation reactions and reductive carbonylation of transition metals. It can also act as catalysts, promoters, halogen/alkoxy or hydroxyl/halogen exchange mediators, reducing agents, and sources of the base.

Application

N,N-Dimethylacetamide is used as a solvent:
  • In the synthesis of poly(amide-hydrazide)s–graphene composites.
  • In the synthesis of N,N′-1,2-ethanediylbis-(4-hydroxy-pentanamide) and 4-hydroxy-N-(2- hydroxyethyl)-pentanamide based polyurethanes in presence of triethylamine catalyst.

Features and Benefits

N,N-Dimethylacetamide has :
  • High dissolution power of various substrate. 
  • High capacity to solvate anions.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.940 - 0.942
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toxic hepatitis in occupational exposure to solvents
Malaguarnera G.et al.
World Journal of Gastroenterology, 18(22), 2756-2756 (2012)
N,N-Dimethylformamide and?N,N-Dimethylacetamide as Carbon, Hydrogen, Nitrogen, and/or Oxygen Sources
Jean LB, et al.
Solvents as Reagents in Organic Synthesis: Reactions and Applications (2017)
Poly (amide-hydrazide) s--graphene composites synthesis and characterization
Elsabee MZ, et al.
Polymer Composites, 38(7), 1311-1318 (2017)
Synthesis and characterization of novel polyurethanes based on N, N'-1, 2-ethanediylbis-(4-hydroxy-pentanamide) and 4-hydroxy-N-(2-hydroxyethyl)-pentanamide
Chalid M
Advanced Materials Research, 277, 112-119 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service