Skip to Content
Merck
All Photos(2)

Key Documents

8.01964

Sigma-Aldrich

1,3-Butanediol

for synthesis

Synonym(s):

(±)-1,3-Butanediol, 1,3-Butylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2CH2OH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1731276
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-529-7
NACRES:
NA.22
grade:
for synthesis
bp:
203-204 °C (lit.)
vapor pressure:
0.06 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

for synthesis

Quality Level

vapor density

3.1 (20 °C, vs air)

vapor pressure

0.06 mmHg ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

741 °F

potency

18610 mg/kg LD50, oral (Rat)
>20000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.9-12.6 % (v/v)

dilution

(for synthesis)

refractive index

n20/D 1.44 (lit.)

pH

6.1 (20 °C, 500 g/L in H2O)

bp

203-204 °C (lit.)

mp

-57 °C

transition temp

flash point 115 °C

solubility

>500 g/L (miscible)

density

1.005 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CC(O)CCO

InChI

1S/C4H10O2/c1-4(6)2-3-5/h4-6H,2-3H2,1H3

InChI key

PUPZLCDOIYMWBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Process design for acidic and alcohol based deep eutectic solvent pretreatment and high pressure homogenization of palm bunches for nanocellulose: Studies DES based on ChCl and 1,3-butanediol, offering insights into process optimizations for clear solution preparation (Sonyeam et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.003 - 1.005
Water (K. F.): ≤ 0.50 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service