Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41122409
NACRES:
NB.61
product line
MVP ICON®
Quality Segment
form
liquid
feature
calibration
manufacturer/tradename
MVP ICON®
application(s)
food and beverages
compatibility
for use with MVP ICON®Conductivity & Concentration Probe (78089BC)
General description
Conductivity Standard Solution (500 μS/cm) for MVP ICON® Conductivity & Concentration Probe
SPECIFICATIONS:
SPECIFICATIONS:
- Potassium chloride conductivity Standard, 500 μS/cm, at 25 °C
- Conductivity at 25 °C: 499 - 501 μS/cm
- Traceable to NIST SRM 3193
Application
For use in performing 3-point calibration of the MVP ICON® pH/Temperature Probe (78088BC)
Packaging
Clear plastic bottle with white screw cap
Legal Information
MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.