Recommended Products
form
liquid
Quality Level
manufacturer/tradename
Novagen®
storage condition
do not freeze
shipped in
wet ice
storage temp.
2-8°C
General description
It comprises a 15-amino acid sequence Lys-Glu-Thr-Ala-Ala-Ala-Lys-Phe-Glu-Arg-Gln-His-Met-Asp-Ser (KETAAAKFERQHMDS).
S-protein Agarose specifically retains S•Tag fusion proteins. Binding can be performed in a column or batch mode. Elution is possible using slightly denaturing conditions or using a specific protease, such as thrombin or enterokinase, with appropriate recombinant proteins. The capacity of S-protein Agarose varies and is based on the size and folding characteristics of a given target protein.
Application
S-protein Agarose has been used in the purification of recombinant S based tagged fusion proteins.
Warning
Toxicity: Standard Handling (A)
Legal Information
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Qian Li et al.
Journal of virology, 93(5) (2018-12-14)
Human herpesvirus 8 (HHV-8) viral interleukin-6 (vIL-6) localizes largely to the endoplasmic reticulum (ER) and here associates functionally with both the gp130 signal transducer and the novel ER membrane protein vitamin K epoxide reductase complex subunit 1 variant-2 (VKORC1v2). The
Toshiki Takahashi et al.
EMBO reports, 20(4) (2019-02-26)
Rab family small GTPases are master regulators of distinct steps of intracellular vesicle trafficking in eukaryotic cells. GDP-bound cytoplasmic forms of Rab proteins are prone to aggregation due to the exposure of hydrophobic groups but the machinery that determines the
Xiaoting Liang et al.
Theranostics, 10(17), 7747-7757 (2020-07-21)
Background: Rab22a-NeoF1 fusion gene containing the 1-38aa of Rab22a (Rab22a1-38) plays a decisive role in driving tumor metastasis by activating RhoA via binding to SmgGDS607. However, its intercellular regulation remains unknown. Methods: The Lys7 (K7) acetylation of Rab22a-NeoF1 was initially
Gadd45a Protein Promotes Skeletal Muscle Atrophy by Forming a Complex with the Protein Kinase MEKK4.
Steven A Bullard et al.
The Journal of biological chemistry, 291(34), 17496-17509 (2016-07-01)
Skeletal muscle atrophy is a serious and highly prevalent condition that remains poorly understood at the molecular level. Previous work found that skeletal muscle atrophy involves an increase in skeletal muscle Gadd45a expression, which is necessary and sufficient for skeletal
Ivette M Sosa Seda et al.
Hepatology research : the official journal of the Japan Society of Hepatology, 40(7), 701-710 (2010-06-19)
Induction of hepatic stellate cell (HSC) apoptosis is a viable therapeutic strategy to reduce liver fibrogenesis. Although BH3-only proteins of the Bcl-2 family trigger pro-apoptotic pathways, the BH3-only proteins mediating HSC apoptosis have not been well defined. Our aim, using
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service