Skip to Content
Merck
All Photos(1)

Key Documents

691400

Sigma-Aldrich

Zebularine

A cell-permeable chemically stable cytidine analog that displays antitumor properties.

Synonym(s):

Zebularine, 1-(β-D-Ribofuranosyl)-1,2-dihydropyrimidin-2-one, 2-Pyrimidone-1-β-D-riboside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O5
CAS Number:
Molecular Weight:
228.20
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 5 mg/mL
methanol: 5 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

N2(C=CC=NC2=O)C1OC(C(C1O)O)CO

InChI

1S/C9H12N2O5/c12-4-5-6(13)7(14)8(16-5)11-3-1-2-10-9(11)15/h1-3,5-8,12-14H,4H2

InChI key

RPQZTTQVRYEKCR-UHFFFAOYSA-N

General description

A cell-permeable chemically stable cytidine analog that displays antitumor properties. Acts as transition state analog inhibitor of cytidine deaminase by binding at the active site as covalent hydrates. Also shown to inhibit DNA methylation and tumor growth both in vitro and in vivo.
A chemically stable, cytidine analog that displays anti-tumor properties. Acts as a transition state analog inhibitor of cytidine deaminase by binding to the active size as covalent hydrates. Also shown to inhibit DNA methylation and tumor growth both in vitro and in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Cytidine deaminase
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

Slight warming may be required to achieve complete solubilization.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.





From Catalog:

Desc. Field- added "cell-permeable"

Other Notes

Cheng, J.C., et al. 2004. Cancer Cell6, 151.
Cheng, J.C., et al. 2003. J. Natl. Cancer Inst.95, 399.
Zhou, L., et al. 2002. J. Mol. Biol.321, 591.
Barchi, J.J., Jr., et al. 1992. J. Org. Chem.57, 536.
Frick, L., et al. 1989. Biochemistry28, 9423.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service