Skip to Content
Merck
All Photos(1)

Key Documents

69050

Sigma-Aldrich

Anti-Cre Antibody

Novagen®

Synonym(s):

Cre Recombinase Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

antibody form

purified antibody

Quality Level

antibody product type

primary antibodies

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Recombinase cre (UniProt: P06956; also known as Cre) is encoded by the cre gene (Gene ID: 2777477) in Escherichia phage P1 (Bacteriophage P1). Cre (Causes recombination with Cyclization recombinase), a 38 kDa DNA recombinase of the ′phage′ integrase family, is derived from the P1 bacteriophage. It catalyzes site-specific recombination and is highly specific for a 34-bp DNA sequence motif called loxP (locus of X-over P1) found in P1 DNA. The loxP site is composed of a variable asymmetric 8-bp sequence in between two sets of 13-bp palindromic sequences (ATAACTTCGTATA-NNNTANNN-TATACGAAGTTAT). Two cre recombinase proteins, each bind to one of the two 13-bp palindromic sequences at a loxP site, form a dimer. This dimer then binds to a second dimer on another loxP site to form a tetramer. The two sites joined by the tetramer are parallel in orientation and the double stranded DNA is cut at both loxP sites. The strands are then rejoined with DNA ligase. Because the Cre gene and loxP sites are not native to most species, they are often introduced into target cells or tissues and employed as a means to precisely control the expression of genes both in cultured cells in vitro and in animals in vivo.
Specific, sensitive detection of Cre recombinase in cells and cell lysates
Supplied as purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine, 0.15 M NaCl, pH 7.4 with 0.05% sodium azide and 50% glycerol.



Evaluated by Western Blotting with Recombinant Cre-His.



Western Blotting Analysis: Representative lot data.Recombinant Cre-His loaded at 20 ng/lane (Lane 1) and 10 ng/lane (Lane 2) was probed with Cat. No. 69050-3, Anti-Cre (1:10,000 dilution). Proteins were visualized using a Donkey Anti-Rabbit secondary antibody conjugated to HRP and a chemiluminescence detection system. Arrow indicates Cre (~39 kDa).

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ken-Ichiro Kuwako et al.
Cell reports, 24(11), 2808-2818 (2018-09-13)
Strictly controlled dendrite patterning underlies precise neural connection. Dendrite self-avoidance is a crucial system preventing self-crossing and clumping of dendrites. Although many cell-surface molecules that regulate self-avoidance have been identified, the signaling pathway that orchestrates it remains poorly understood, particularly in mammals. Here
Feng Li et al.
Frontiers in genetics, 3, 255-255 (2013-01-08)
The tongue epithelium is one of the most rapidly self-renewing tissues in adult mammals. Multiple stem cell populations are currently believed to exist in tongue epithelia. Keratin 14 (K14) positive cells differentiate into either lingual epithelia or lingual papillae, while
Fei Gao et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(32), 11987-11992 (2006-08-01)
Mutation of the transcription factor and tumor suppressor gene WT1 results in a range of genitourinary anomalies in humans, including 46,XY gonadal dysgenesis, indicating that WT1 plays a critical role in sex determination. However, because knockout of Wt1 in mice
Yang Shi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(25), 8129-8142 (2009-06-26)
Dendritic spines are the postsynaptic sites of most excitatory synapses in the brain and are highly enriched in polymerized F-actin, which drives the formation and maintenance of mature dendritic spines and synapses. We propose that suppressing the activity of the
Amanda M Ackermann et al.
Cell metabolism, 28(5), 787-792 (2018-07-31)
Recent reports identified activation of the GABA signaling pathway as a means to induce transdifferentiation of pancreatic α cells into β cells. These reports followed several previous studies that found that α cells were particularly well suited to conversion into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service