Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

655203

Sigma-Aldrich

TX-1918

A cell-permeable arylidene-cyclopentenedione derived tyrphostin that acts as a potent inhibitor for eEF2-K, while it inhibits other kinases at much higher concentrations.

Synonym(s):

TX-1918, 2-((3,5-dimethyl-4-hydroxyphenyl)-methylene)-4-cyclopentene-1,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O3
CAS Number:
Molecular Weight:
228.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light orange

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Oc1c(cc(cc1C)C=C2C(=O)C=CC2=O)C

General description

A cell-permeable arylidene-cyclopentenedione derived tyrphostin that acts as a potent inhibitor for eEF2-K (IC50 = 440 nM), while it inhibits other kinases at much higher concentrations (IC50 = 4.4, 44, 44, and 440 µM for Src, PKA, PKC, and EGFR-K, respectively). Displays potent antitumor activity (EC50 = 2.07 and 230 µM in HepG2 and HCT116 tumor cells, respectively) with greatly reduced mitochondrial- (≥5000-fold) and hepato-toxicity (≥90-fold) when compared with another tyrphostin, AG 17 (Cat. No. 658425). It is therefore a more promising candidate as a therapeutic agent for cancer treatment.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
eEF2-K
Product does not compete with ATP.
Reversible: no
Target IC50: 440 nM against eEF2-K

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Hori, H., et al. 2002. Bioorg. Med. Chem.10, 3257.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service