Skip to Content
Merck
All Photos(1)

Key Documents

572635

Sigma-Aldrich

SU6656

A potent, cell-permeable, reversible, and ATP-competitive Src family kinase inhibitor.

Synonym(s):

SU6656

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21N3O3S
CAS Number:
Molecular Weight:
371.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

burnt orange

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[S](=O)(=O)(N(C)C)c1cc2c(cc1)NC(=O)\C\2=C\c3[nH]c4c(c3)CCCC4

InChI

1S/C19H21N3O3S/c1-22(2)26(24,25)14-7-8-18-15(11-14)16(19(23)21-18)10-13-9-12-5-3-4-6-17(12)20-13/h7-11,20H,3-6H2,1-2H3,(H,21,23)/b16-10+

InChI key

LOGJQOUIVKBFGH-MHWRWJLKSA-N

General description

A potent, cell-permeable, reversible, and ATP-competitive Src family kinase inhibitor. Inhibits Src (IC50 = 280 nM) as well as closely related kinases such as Fyn (IC50 = 170 nM) and Yes (IC50 = 20 nM) and Lyn (IC50 = 130 nM). Acts as a weak inhibitor of Lck (IC50 = 688 µM) and PDGF receptor kinase (IC50 >10 µM). A 10 mM (500 µg/135 µl) solution of SU6656 (Cat. No. 572636) in DMSO is also available.
A potent, cell-permeable, reversible, and ATP-competitive inhibitor of Src-family protein tyrosine kinases. Inhibits Src (IC50 = 280 nM) as well as closely related kinases Fyn (IC50 = 170 nM), Yes (IC50 = 20 nM) and Lyn (IC50 = 130 nM). The inhibition is competitive with respect to ATP. Has only a trivial effect on Lck (IC50 = 6.88 µM). Does not affect the activity of PDGF-β receptor kinase (IC50 ≥10 µM) and IGF-1 receptor kinase (IC50 ≥20 µM).

Biochem/physiol Actions

Cell permeable: yes
Product competes with ATP.
Reversible: yes
Target IC50: 280 nM, 170 nM, 20 nM, and 130 nM against Src, Fyn, Yes, and Lyn.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Bowman, T., et al. 2001. Proc. Natl. Acad. Sci. USA98, 7319.
Blake, R.A., et al. 2000. Mol. Cell. Biol.20, 9018.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Wu et al.
Cell reports, 30(4), 1129-1140 (2020-01-30)
Plasma membrane damage and cell death during processes such as necroptosis and apoptosis result from cues originating intracellularly. However, death caused by pore-forming agents, like bacterial toxins or complement, is due to direct external injury to the plasma membrane. To

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service