Skip to Content
Merck
All Photos(1)

Key Documents

5.33851

Sigma-Aldrich

Cancer Stemness Inhibitor, BBI608

Synonym(s):

Cancer Stemness Inhibitor, BBI608, 2-Acetyl-4H,9H-naphtho[2,3-b]furan-4,9-dione, 2-Acetylnaphtho[2,3-b]furan-4,9-dione, Spherogenesis Blocker, BBI-608, Napabucasin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8O4
CAS Number:
Molecular Weight:
240.21
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Pricing and availability is not currently available.

Assay

≥97% (HPLC)

Quality Level

form

solid

potency

142 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

[o]1c2c(cc1C(=O)C)C(=O)c3c(cccc3)C2=O

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HPA024756HPA040134HPA053874
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

A cell-permeable, bioactive, non-toxic naphthofurandione analog that blocks STAT3-driven gene transcription. Selectively diminishes gene expression and self-renewal in stemness high cancer cells. Shown to be more effective in stemness high cancer cell population (IC50 = 142 nM) compared to regular cancer cells (IC50 = 395 nM). Reduces spherogenesis in CD44 expressing cancer cells without affecting the colony formation in CD34+ hematopoietic stem cells. Effectively reduces the levels of Nanog, Axl, Sox-2, Klf4, survivin, c-Myc, Bmi-1, and β-catenin within a few hours following treatment. Displays broad spectrum activity against stemness high cancer cells from multiple tumors (IC50 = 291, 432, 624, 479, 549, and 729 nM for CAOV-3, SW-626, PaCa2, SNU-475, H1975, and U87-MG, respectively). Suppresses the growth of PaCa-2 xenografts in murine models that exhibited relapse of tumor growth (20 mg/kg, i.p, q.d.) following the cessation of 41-day gemcitabine treatment. Also shown to block cancer relapse, and spleen and liver metastasis in the intrasplenic nude mouse model injected with HT29 colon cancer cells.
A cell-permeable, bioactive, non-toxic naphthofurandione analog that blocks STAT3-driven gene transcription. Selectively diminishes gene expression and self-renewal in stemness high cancer cells. Shown to be more effective in stemness high cancer cell population (IC50 = 142 nM) compared to regular cancer cells (IC50 = 395 nM). Reduces spherogenesis in CD44 expressing cancer cells without affecting the colony formation in CD34+ hematopoietic stem cells. Effectively reduces the levels of Nanog, Axl, Sox-2, Klf4, survivin, c-Myc, Bmi-1, and β-catenin within a few hours following treatment. Displays broad spectrum activity against stemness high cancer cells from multiple tumors (IC50 = 291, 432, 624, 479, 549, and 729 nM for CAOV-3, SW-626, PaCa2, SNU-475, H1975, and U87-MG, respectively). Suppresses the growth of PaCa-2 xenografts in murine models that exhibited relapse of tumor growth (20 mg/kg, i.p, q.d.) following the cessation of 41-day gemcitabine treatment. Also shown to block cancer relapse, and spleen and liver metastasis in the intrasplenic nude mouse model injected with HT29 colon cancer cells.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
STAT3

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Li, Y., et al. 2015. Proc. Natl. Acad. Sci. USA112, 1839.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service