Skip to Content
Merck
All Photos(1)

Key Documents

5.30535

Sigma-Aldrich

RPA Inhibitor, TDRL-505

Synonym(s):

RPA Inhibitor, TDRL-505, Replication Protein A Inhibitor, Pot1 Inhibitor, Protection of Telemeres Protein 1 Inhibitor, TDRL505

Sign Into View Organizational & Contract Pricing

Select a Size

25MG
CA$37.90

CA$37.90


Please contact Customer Service for Availability

To order products, please contact your local dealer.

Select a Size

Change View
25MG
CA$37.90

About This Item

Empirical Formula (Hill Notation):
C24H21BrClN3O4
CAS Number:
Molecular Weight:
530.80
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

CA$37.90


Please contact Customer Service for Availability

To order products, please contact your local dealer.

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

SMILES string

CCOC1=CC2=NC(=C(C=C2C=C1)C3CC(=NN3C(=O)CCC(=O)O)C4=CC=C(C=C4)Br)Cl

General description

A cell-permeable quinolinyl-dihydropyrazolyl-oxobutanoic acid compound that is shown to target human Replication Protein A (RPA) via primarily the oligosaccharide/oligonucleotide-binding (OB) folds of DNA-bonding domains A & B (DBD-A/B) in the p70 subunit, effectively inhibiting its DNA-binding activity as well as that of Schizosaccharomyces pombe telemere end-binding protein Pot1 (IC50= 12.9 µM and 15.7 µM, respectively), but not Xeroderma pigmentosum group A protein (XPA) involved in the nucleotide excision repair (NER) pathway or E. coli SSB. Reported to induce cell cycle G1-arrest and decrease Annvex V-/PI- healthy cell population (by 50%; 30.8 µM for 48 h) in H460 non-small cell lung carcinoma (NSCLC) cultures with minimal cytotoxicity against freshly isolated human PBMC. Potentiates cisplatin (Cat. No. 232120) and etopside (Cat. No. 341205) cytotoxicity in H460 cultures in a synergistic manner.
A cell-permeable quinolinyl-dihydropyrazolyl-oxobutanoic acid compound that is shown to target human Replication Protein A via primarily the oligosaccharide/oligonucleotide-binding (OB) folds of DNA-bonding domains A & B (DBD-A/B) in the p70 subunit, effectively inhibiting its DNA-binding activity as well as that of Schizosaccharomyces pombe telemere end-binding protein Pot1 (IC50= 12.9 µM/25 nM hRPA/25 nM 34-base AG-rich ssDNA and 15.7 µM/20 nM SpPot1 DBD/25 nM 15-base ssDNA), but not Xeroderma pigmentosum group A protein (XPA) involved in the nucleotide excision repair (NER) pathway or E. coli SSB (20% inhibition at 100 µM against 3.3 nM EcSSB & 25 nM 3Pc3 ssDNA binding). Reported to cause decreased nuclear RPA staining (50 µM for 3 h), cell cycle G1-arrest (75 to 100 µM), and a drop in Annvex V-/PI- healthy cell population (by 50%; 30.8 µM for 48 h) in H460 non-small cell lung carcinoma (NSCLC) cultures with minimal cytotoxicity against freshly isolated human PBMC. Potentiates cisplatin (Cat. No. 232120) and etopside (Cat. No. 341205) cytotoxicity in H460 cultures in a synergistic manner, consistent with blockage of RPA-mediated NER pathway.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Replication Protein A
Reversible: yes
Target IC50: 12.9, 15.7 and 30.8 &micro

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Use only fresh DMSO for reconstitution.

Other Notes

Anciano Granadillo, V.J., et al. 2010. J. Nucleic Acids2010, 304035.

Shuck, S.C., and Turchi, J.J. 2010. Cancer Res.70, 3189.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service