Skip to Content
Merck
All Photos(1)

Key Documents

517301

Sigma-Aldrich

P-Glycoprotein Inhibitor, C-4

The P-Glycoprotein Inhibitor, C-4 controls the biological activity of P-Glycoprotein. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

P-Glycoprotein Inhibitor, C-4, (E)-4-Chloro-N-(3-(3-(4-hydroxy-3-methoxyphenyl)acryloyl)phenyl)benzamide, C-4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H18ClNO4
CAS Number:
Molecular Weight:
407.85
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

COC1=C(C=CC(=C1)C=CC(=O)C2=CC(=CC=C2)NC(=O)C3=CC=C(C=C3)Cl)O

InChI

1S/C23H18ClNO4/c1-29-22-13-15(6-12-21(22)27)5-11-20(26)17-3-2-4-19(14-17)25-23(28)16-7-9-18(24)10-8-16/h2-14,27H,1H3,(H,25,28)

General description

A cell-permeable cinnamoyl compound that reversibly inhibits P-gp (P-glycoprotein) efflux function and reverses the MDR (multidrug resistance) phenotype with no noticeable effect on P-gp expression. Shown to significantly enhance Paclitaxel- (Cat. No. 580555) and Vincristine- (Cat. No. 677181) induced cytotoxicity as well as apoptosis in P-gp expressing KBV20C cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
P-Glycoprotein
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Kim, Y.K., et al. 2007. Biochem. Biophys. Res. Commun.355, 136.
Woo, H.B., et al. 2005. Bioorg. Med. Chem. Lett.15, 3782.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service