Skip to Content
Merck
All Photos(1)

Key Documents

5.04907

Sigma-Aldrich

Tankyrase 1/2 Inhibitor VI, G007-LK

Synonym(s):

Tankyrase 1/2 Inhibitor VI, G007-LK, TNKS1/2 Inhibitor VI, G007LK, Wnt Pathway Inhibitor XXI, 4-(5-(( E)-2-(4-(2-Chlorophenyl)-5-(5-(methylsulfonyl)pyridin-2-yl)-4H-1,2,4-triazol-3-yl)ethenyl)-1,3,4-oxadiazol-2-yl)benzonitrile, TNKS1/2 Inhibitor VI, G007LK, Wnt Pathway Inhibitor XXI, 4-(5-((E)-2-(4-(2-Chlorophenyl)-5-(5-(methylsulfonyl)pyridin-2-yl)-4H-1,2,4-triazol-3-yl)ethenyl)-1,3,4-oxadiazol-2-yl)benzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H16ClN7O3S
CAS Number:
Molecular Weight:
529.96
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(C)c1cnc(cc1)c2[n](c(nn2)\C=C\c4nnc([o]4)c5ccc(cc5)C#N)c3c(cccc3)Cl

InChI

1S/C25H16ClN7O3S/c1-37(34,35)18-10-11-20(28-15-18)24-31-29-22(33(24)21-5-3-2-4-19(21)26)12-13-23-30-32-25(36-23)17-8-6-16(14-27)7-9-17/h2-13,15H,1H3/b13-12+

General description

A cell-permeable, triazolyl-vinyl-oxadiazole compound that targets adenosine site of the NAD+ pocket in the TNKS PARP domain and acts as a potent and highly selective tankyrase inhibitor (IC50 = 33 and 26 nM, respectively, against TNKS1/PARP5a/ARTD5 and TNKS2/PARP5b/ARTD6 in auto-PARsylation assays), effectively inhibiting murine Wnt3a-induced reporter activity in human HEK293 and murine 10T1/2 cultures (ICmax ~300 and 600 nM, respectively). Shown to display little or no inhibitory potency (IC50 >10 µM) toward 7 other PARP enzymes (PARP1, 2, 3, 6, 7,10,11), 90 kinases, 16 phosphatases, and 75 GPCRs (IC50 >10 µM). Shown to inhibit two CRC lines, COLO-320DM & SW403, colony formation in vitro (200 nM) and tumor expansion in mice in vivo (20 mg/kg via daily i.p.), although G007-LK toxicity is observed at higher dosages (≥30 mg/kg/12 h or 60 mg/kg/d via i.p.).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
A cell-permeable, triazolyl-vinyl-oxadiazole compound that targets adenosine site of the NAD+ pocket in the TNKS PARP domain and acts as a potent and highly selective tankyrase inhibitor (IC50 = 33 and 26 nM, respectively, in TNKS1/PARP5a/ARTD5 and TNKS2/PARP5b/ARTD6 auto-PARsylation assays), effectively inhibiting murine Wnt3a-induced reporter activity in human HEK293 and murine 10T1/2 cultures (ICmax ~300 and 600 nM, respectively). In addition to being more selective over PARP1/2 (IC50 >10 µM vs 116 nM/47 nM with XAV939) than the nicotinamide site-targeting XAV939 (Cat. No. 575545), G007-LK is also shown to display little or no inhibitory potency (IC50 >10 µM) toward 5 other PARP enzymes (PARP3, 6, 7,10,11), 90 kinases, 16 phosphatases, and 75 GPCRs (IC50 >10 µM). Wnt signaling inhibition upon G007-LK treatment in 11 APC (Adenomatous polyposis coli/deleted in polyposis 2.5/DP2.5) mutatant CRC (colorectal cancer) lines ranges from no effect to varying degrees of partial blockages that do not correlate with the observed CRC colony formation inhibitions.Two CRC lines, COLO-320DM & SW403, whose colony formation are affected by G007-LK in vitro (by 70% and 27%, respectively, at 200 nM in 7 to 13 d) are also inhibited by G007-LK for their tumor expansion in mice in vivo (by 47% and 42%, respectively, on d21; 20 mg/kg per daily i.p.), while G007-LK toxicity is observed at high dosages in mice (≥30 mg/kg/12 h or 60 mg/kg/d via i.p.) due to small intestine damage as a result of on-target wnt signaling inhibition in normal tissue. Exhibits favorable pharmacokinetic properties in mice when administered via i.p. ( Plasma t1/2 ≥2.73 h; Cmax ≥2.358 µg/mL; 5 mg/kg) or p.o. ( Plasma t1/2 ≥2.8 h; Cmax ≥871 ng/mL; 5 mg/kg).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Tankyrase 1/2
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Lau, T., et al. 2013. Cancer Res.73, 3132.
Voronkov, A., et al. 2013. J. Med. Chem.56, 3012.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service