Skip to Content
Merck
All Photos(1)

Key Documents

496100

Sigma-Aldrich

Omeprazole

A cell-permeable pyridyl methylsulfinyl benzimidazole compound that acts as selective proton pump inhibitor.

Synonym(s):

Omeprazole, H 168/68, 5-Methoxy-2[(4-methoxy-3,5-dimethyl-2-pyridyl)methylsulfinyl]-1H-benzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N3O3S
CAS Number:
Molecular Weight:
345.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

description

Merck USA index - 14, 6845

Assay

≥94% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan to off-white

solubility

DMSO: 25 mg/mL

cation traces

heavy metals: ≤20 ppm

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S+]([O-])(Cc3ncc(c(c3C)OC)C)c1[nH]c2c(n1)cc(cc2)OC

InChI

1S/C17H19N3O3S/c1-10-8-18-15(11(2)16(10)23-4)9-24(21)17-19-13-6-5-12(22-3)7-14(13)20-17/h5-8H,9H2,1-4H3,(H,19,20)

InChI key

SUBDBMMJDZJVOS-UHFFFAOYSA-N

General description

A cell-permeable pyridyl methylsulfinyl benzimidazole compound that acts as selective proton pump inhibitor. Behaves as a prodrug by undergoing an acid-catalyzed rearrangement to a thiol-reactive cationic sulfenamide that inhibits (H+, K+)-ATPase in the gastric milieu. An aryl hydrocarbon-like inducer of cytochrome P450 secretion in human liver.
A cell-permeable, selective proton pump inhibitor. Behaves as a prodrug by undergoing an acid-catalyzed rearrangement to a thiol-reactive cationic sulfenamide that inhibits (H+-K+)-ATPase in the gastric milieu. Also acts as an aryl hydrocarbon-like inducer of cytochrome P450 secretion in human liver.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
proton pump
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lambrecht, N., et al. 2000. J. Biol. Chem.275, 4041.
Besancon, M., et al. 1997. J. Biol. Chem.272, 22438.
Sachs, G., et al. 1995. Ann. Rev. Pharmacol. Toxicol.35, 277.
Diaz, D., et al. 1990. Gastroenterology99, 737.
Fellenius, E., et al. 1981. Nature290, 159.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Sachs et al.
Annual review of pharmacology and toxicology, 35, 277-305 (1995-01-01)
The gastric H+,K+ ATPase--the gastric acid pump--is the molecular target for the class of antisecretory drugs called the proton-pump inhibitors (PPIs). These compounds--omeprazole, lansoprazole, and pantoprazole--contain, as their core structure, 2-pyridyl methylsulfinyl benzimidazole. The H+,K+ ATPase is a heterodimer composed
M Besancon et al.
The Journal of biological chemistry, 272(36), 22438-22446 (1997-09-05)
The vesicular gastric H,K-ATPase catalyzes an electroneutral H for K exchange allowing acidification of the intravesicular space. There is a total of 28 cysteines present in the alpha subunit of the gastric H,K-ATPase, of which 10 are found in the
N Lambrecht et al.
The Journal of biological chemistry, 275(6), 4041-4048 (2000-02-08)
The gastric H,K-ATPase is covalently inhibited by substituted pyridyl-methylsulfinyl-benzimidazoles, such as omeprazole, that convert to thiophilic probes of luminally accessible cysteines in the acid space. The K(+) competitive inhibitor, SCH28080, prevented inhibition of acid transport by omeprazole. In stably expressing
D Diaz et al.
Gastroenterology, 99(3), 737-747 (1990-09-01)
Omeprazole is a new drug used for its high efficiency as an inhibitor of gastric acid secretion. This substituted benzimidazole molecule had been shown to decrease several liver cytochrome P450-mediated monooxygenase activities both in vitro and in vivo. The present
E Fellenius et al.
Nature, 290(5802), 159-161 (1981-03-12)
Studies both in vivo and in vitro have shown that substituted benzimidazoles inhibit the stimulation of acid secretion produced by dibutyryl cyclic AMP and histamine. Furthermore, the results differ from those produced by H2 antagonists and anticholinergic agents in that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service