Skip to Content
Merck
All Photos(1)

Key Documents

492018

Millipore

NP-40 Alternative, PROTEIN GRADE® Detergent, 10% Solution, Sterile-Filtered

A non-ionic surfactant useful for the isolation and purification of functional membrane protein complexes. This detergent has been purified to reduce levels of contaminating aldehydes, metals, peroxides, and salts.

Synonym(s):

Nonylphenyl Polyethylene Glycol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

description

RTECS - MD0905000

form

solution

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤200 μM Aldehyde
≤200 μM peroxide

color

clear colorless

conductivity

≤50 μmho

CMC

50 - 300 μM

shipped in

ambient

storage temp.

2-8°C

SMILES string

O(CCOc1ccc(cc1)CCCCCCCCC)CCO

InChI

1S/C19H32O3/c1-2-3-4-5-6-7-8-9-18-10-12-19(13-11-18)22-17-16-21-15-14-20/h10-13,20H,2-9,14-17H2,1H3

InChI key

BLXVTZPGEOGTGG-UHFFFAOYSA-N

General description

A non-ionic surfactant used in the isolation and purification of functional membrane protein complexes and in two-dimensional gel electrophoresis (2DGE). Similar in usage to NP-40 and TRITON X-100 (Cat. No. 648462). Purified to reduce levels of contaminating aldehydes, metals, peroxides, and salts. CMC: 50-300 µM.
A non-ionic surfactant useful for the isolation and purification of functional membrane protein complexes. This detergent has been purified to reduce levels of contaminating aldehydes, metals, peroxides, and salts.

Warning

Toxicity: Irritant (B)

Physical form

10% aqueous solution.

Reconstitution

Following initial use, aliquot and freeze (-20°C).

Other Notes

Ripoche, P., et al. 1982. Biochim. Biophys. Acta693, 497.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
PROTEIN GRADE is a registered trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsung-Han S Hsieh et al.
Molecular cell, 78(3), 539-553 (2020-03-28)
Whereas folding of genomes at the large scale of epigenomic compartments and topologically associating domains (TADs) is now relatively well understood, how chromatin is folded at finer scales remains largely unexplored in mammals. Here, we overcome some limitations of conventional

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service