Skip to Content
Merck
All Photos(1)

Key Documents

484235

Sigma-Aldrich

p-Nitroblue Tetrazolium Chloride

≥98% (TLC), crystalline solid, nitric oxide synthase inhibitor, Calbiochem®

Synonym(s):

p-Nitroblue Tetrazolium Chloride, NBT, Nitro BT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H30Cl2N10O6
CAS Number:
Molecular Weight:
817.64
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

p-Nitroblue Tetrazolium Chloride, NADPH-diaphorase substrate that competitively inhibits nitric oxide synthase (IC₅₀ = 3-4 µM).

Quality Level

description

RTECS - XF8045000

Assay

≥98% (TLC)

form

crystalline solid

potency

3-4 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

methanol: soluble
water: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].[n+]2([n](nc(n2)c8ccccc8)c3c(cc(cc3)c4cc(c(cc4)[n]5[n+](nc(n5)c7ccccc7)c6ccc(cc6)[N+](=O)[O-])OC)OC)c1ccc(cc1)[N+](=O)[O-]

InChI

1S/C40H30N10O6.2ClH/c1-55-37-25-29(13-23-35(37)47-43-39(27-9-5-3-6-10-27)41-45(47)31-15-19-33(20-16-31)49(51)52)30-14-24-36(38(26-30)56-2)48-44-40(28-11-7-4-8-12-28)42-46(48)32-17-21-34(22-18-32)50(53)54;;/h3-26H,1-2H3;2*1H/q+2;;/p-2

InChI key

FSVCQIDHPKZJSO-UHFFFAOYSA-L

General description

NADPH-diaphorase substrate that competitively inhibits nitric oxide synthase (IC50 = 3-4 µM). A well-known scavenger of superoxide anions. Useful as a substrate for alkaline phosphatase, often in conjunction with BCIP (Cat. No. 203788).

Biochem/physiol Actions

Primary Target
NOS

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long-term storage or refrigerate (4°C) for short-term storage.

Other Notes

Pistelli, A., et al. 1994. Biochem. Pharmacol. 47, 1737.
De la Fuente, M., et al. 1993. Comp. Immunol. Microbiol. Infect. Dis.16, 29.
Libon, C., et al. 1993. J. Leukoc. Biol.53, 93.
Mittal, C.K. 1993. Biochem. Biophys. Res. Commun. 193, 126.
Nichols, K., et al. 1992. Neuroscience51, 791.
Nichols, K., et al. 1992. Neuroscience51, 791.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Xin Diao et al.
Chinese journal of integrative medicine, 26(10), 736-744 (2019-11-27)
To investigate the phenolic composition, antioxidant properties, and hepatoprotective mechanisms of polyphenols from green tea extract (GTP) in carbon tetrachloride (CCl4)-induced acute liver injury mouse model. High-performance liquid chromatography was used to analyze the chemical composition of the extract. Antioxidant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service