Skip to Content
Merck
All Photos(1)

Key Documents

437644

Sigma-Aldrich

Lipoproteins, Low Density, Human Plasma

Synonym(s):

LDL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

Assay

≥95% (of total lipoprotein content, electrophoresis)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

do not freeze
protect from light

shipped in

wet ice

storage temp.

2-8°C

General description

A 10 mg vial contains ~2 mg of protein.
Native low density lipoproteins from human plasma. Cholesterol-carrier lipoprotein responsible for delivery of lipids (cholesterol) from liver to tissues. (Composition: 78-81% lipid; 19-22% protein. Note: 10 mg vial contains 2 mg of protein.)
Native low density lipoproteins from human plasma. Cholesterol-carrier lipoprotein responsible for delivery of lipids (cholesterol) from liver to tissues. Composition: 78-81% lipid; 19-22% protein.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 0.01% EDTA, pH 7.4.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

This product does not contain preservatives, so it is recommended that it be used under a laminar flow hood. Following initial use, this product is stable for 1 week at 2-8°C.

Other Notes

Foster, J.D., et al. 1993. Endocrinology132, 337.
Retsky, K.L., et al. 1993. J. Biol. Chem. 268, 1304.
Fielding, C.J. 1992. FASEB J. 6, 3162.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiawei Chen et al.
The Biochemical journal, 393(Pt 1), 255-265 (2005-09-22)
LOX-1, a receptor for ox-LDL (oxidized low-density lipoprotein), has recently been determined to play a critical role in the progression of atherosclerosis. LOX-1 expression (mRNA and protein) has been shown to be up-regulated by pro-atherogenic stimuli, such as ox-LDL and
Eui-Jung Park et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(2), 3379-3398 (2020-01-11)
Aquaporin-5 (AQP5) plays a role in breast cancer cell migration. This study aimed to identify AQP5-targeting miRNAs and examine their effects on breast cancer cell migration through exosome-mediated delivery. Bioinformatic analyses identified miR-1226-3p, miR-19a-3p, and miR-19b-3p as putative regulators of
Se-Hyun Oh et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(6), 7301-7314 (2019-03-13)
Hypercholesterolemia is reported to increase reactive oxygen species (ROS) and to promote breast cancer progression. ROS play an important role in tumor biology, and xanthine oxidase (XO) is an enzyme that generates ROS. The effects of febuxostat (FBX), an XO
Yaw Asare et al.
Frontiers in physiology, 11, 673-673 (2020-07-07)
Atherogenesis and arterial remodeling following mechanical injury are driven by inflammation and mononuclear cell infiltration. The binding of immune complexes (ICs) to immunoglobulin (Ig)-Fc gamma receptors (FcγRs) on most innate and adaptive immune cells induces a variety of inflammatory responses
Brett M Stevens et al.
STAR protocols, 2(1), 100248-100248 (2021-01-14)
Isolation of leukemia stem cells presents a challenge due to the heterogeneity of the immunophenotypic markers commonly used to identify blood stem cells. Several studies have reported that relative levels of reactive oxygen species (ROS) can be used to enrich

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service