Skip to Content
Merck
All Photos(1)

Key Documents

428150

Sigma-Aldrich

Lavendustin A

A cell-permeable, reversible, and substrate competitive inhibitor of EGF receptor tyrosine kinase and p60c-src with little effect on protein kinase A or protein kinase C.

Synonym(s):

Lavendustin A, 5-Amino-[(N-2,5-dihydroxybenzyl)-Nʹ-2-hydroxybenzyl]salicylic Acid, RG14355

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19NO6
CAS Number:
Molecular Weight:
381.38
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

ethanol: 10 mg/mL
DMSO: soluble

shipped in

ambient

storage temp.

−20°C

InChI

1S/C21H19NO6/c23-16-6-8-19(25)14(9-16)12-22(11-13-3-1-2-4-18(13)24)15-5-7-20(26)17(10-15)21(27)28/h1-10,23-26H,11-12H2,(H,27,28)

InChI key

ULTTYPMRMMDONC-UHFFFAOYSA-N

General description

A cell-permeable, reversible, and substrate competitive inhibitor of EGF receptor tyrosine kinase (IC50 = 11 nM) and p60c-src (IC50 = 500 nM) with little effect on protein kinase A or protein kinase C (IC50 >200 µM). Also inhibits NMDA-stimulated cGMP production (IC50 = 30 nM). Exhibits anti-proliferative properties. Suppresses the angiogenic action of vascular endothelial growth factor (VEGF) in rats.
A cell-permeable, reversible, substrate competitive, and potent inhibitor of EGF receptor tyrosine kinase (IC50 = 11 nM) and pp60c-src (IC50 = 500 nM) with little effect on protein kinase A or protein kinase C (IC50 >200 µM). Also inhibits NMDA-stimulated cGMP production (IC50 = 30 nM). Exhibits anti-proliferative properties. Suppresses the angiogenic action of vascular endothelial growth factor in rats.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
EGF receptor tyrosine kinase
Product does not compete with ATP.
Reversible: yes
Target IC50: 11 nM and 500 nM against EGF receptor tyrosine kinase and p60c-src, respectively

Warning

Toxicity: Standard Handling (A)

Preparation Note

Further dilute with aqueous buffer just prior to use.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Hu, D.E., and Fan, T.-P. 1995. Br. J. Pharmacol. 114, 262.
Rodriguez, J., et al. 1994. Neuropharmacology 33, 1267.
Hsu, C.-Y., et al. 1991. J. Biol. Chem.266, 21105.
O’Dell, T.J., et al. 1991. Nature 353, 558; Onoda, T., et al. 1989. J. Nat. Prod.52, 1252.
Bishop, J.M. 1983. Annu. Rev. Biochem. 52, 307.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Rodriguez et al.
Neuropharmacology, 33(11), 1267-1274 (1994-11-01)
In striatal neurones in culture, N-methyl-D-aspartate-(NMDA), kainate-(Kai) and K(+)-dependent cGMP production is entirely mediated via nitric oxide (NO). Low concentrations of lavendustin-A (< or = 0.3 microM), a highly specific tyrosine kinase inhibitor, reduced irreversibly and in a time-dependent manner
Cellular oncogenes and retroviruses.
J M Bishop
Annual review of biochemistry, 52, 301-354 (1983-01-01)
C Y Hsu et al.
The Journal of biological chemistry, 266(31), 21105-21112 (1991-11-05)
Lavendustin-A was reported to be a potent tyrosine kinase inhibitor of the epidermal growth factor (EGF) receptor (Onoda, T., Iinuma, H., Sasaki, Y., Hamada, M., Isshibi, K., Naganawa, H., Takeuchi, T., Tatsuta, K., and Umezawa, K. (1989) J. Nat. Prod.
D E Hu et al.
British journal of pharmacology, 114(2), 262-268 (1995-01-01)
1. Vascular endothelial growth factor (VEGF) is a heparin-binding angiogenic factor which specifically acts on endothelial cells via distinct membrane-spanning tyrosine kinase receptors. Here we used the rat sponge implant model to test the hypothesis that the angiogenic activity of
T J O'Dell et al.
Nature, 353(6344), 558-560 (1991-10-10)
Long-term potentiation (LTP) in the hippocampus is thought to contribute to memory formation. In the Ca1 region, LTP requires the NMDA (N-methyl-D-aspartate) receptor-dependent influx of Ca2+ and activation of serine and threonine protein kinases. Because of the high amount of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service