Skip to Content
Merck
All Photos(1)

Key Documents

428029

Millipore

Sodium dodecyl sulfate

Tablets, An ionic detergent useful in electrophoretic separation of proteins and lipids.

Synonym(s):

SDS Tablets, Sodium Dodecyl Sulfate Tablets

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H25O4S · Na
CAS Number:
Molecular Weight:
288.38
MDL number:
UNSPSC Code:
12352200

Quality Level

mol wt

micellar wt 18,000

packaging

pkg of 50 tablets

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

aggregation number

62

CMC

7-10 mM

shipped in

ambient

storage temp.

15-25°C

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

General description

An ionic detergent useful in electrophoretic separation of proteins and lipids.
An ionic detergent useful in electrophoretic separation of proteins and lipids. Aggregation number: 62; CMC: 7-10 mM; micellar weight: 18,000.

Warning

Toxicity: Flammable (J)

Physical form

Each tablet contains 0.5 g SDS.

Preparation Note

Dissolve one tablet in the desired volume of H₂O.

Reconstitution

Following reconstitution, store at room temperature. Stock solutions are stable for up to 6 months at room temperature.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service