Skip to Content
Merck
All Photos(2)

Key Documents

420358

Sigma-Aldrich

CBFβ Activator, Kartogenin

The CBFβ Activator, Kartogenin modulates the biological activity of CBFβ. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

CBFβ Activator, Kartogenin, 2-(Biphenyl-4-ylcarbamoyl)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H15NO3
CAS Number:
Molecular Weight:
317.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N(c2ccc(cc2)c3ccccc3)C(=O)c1c(cccc1)C(=O)O

InChI

1S/C20H15NO3/c22-19(17-8-4-5-9-18(17)20(23)24)21-16-12-10-15(11-13-16)14-6-2-1-3-7-14/h1-13H,(H,21,22)(H,23,24)

General description

A cell-permeable biphenylcarbamoylbenzoate compound that potently induces the differentiation of human mesenchymal stem cells (hMSCs) into chondrocytes (EC50 = 100 nM). Reversibly binds to the FC-1 fragment of filamin A and disrupts its association with CBFβ leading to nuclear localization of CBFβ and binding to runt-related transcription factor (RUNX) to regulate chondrogenesis. Also reported to inhibit cytokine-induced release of nitric oxide (NO) and glycosaminoglycan (GAG). Exhibits chondroprotective properties and promotes cartilage repair in murine osteoarthritis models. Shown to up-regulate the levels of TIMP1, collagen, and aggrecan thereby blocking further matrix breakdown without affecting the activities of MMP-3, MMP-13, and aggrecanase in primary chondrocytes or hMSCs.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Johnson, K., et al. 2012. Science336, 717.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service