Skip to Content
Merck
All Photos(1)

Key Documents

390238

Sigma-Aldrich

Camptothecin

from Camptotheca acuminata, ≥95% (HPLC), powder, DNA topoisomerase I inhibitor, Calbiochem®

Synonym(s):

Camptothecin, 10-Hydroxy-, Camptotheca acuminata, 10-Hydroxycamptothecin, HCPT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16N2O5
CAS Number:
Molecular Weight:
364.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Camptothecin, 10-Hydroxy-, Camptotheca acuminata, A cell-permeable powerful DNA topoisomerase I inhibitor.

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

pale yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]21c(cc5c([c]2=O)COC(=O)C5(O)CC)c3nc4c(cc3C1)cc(cc4)O

InChI

1S/C20H16N2O5/c1-2-20(26)14-7-16-17-11(5-10-6-12(23)3-4-15(10)21-17)8-22(16)18(24)13(14)9-27-19(20)25/h3-7,23,26H,2,8-9H2,1H3

InChI key

HAWSQZCWOQZXHI-UHFFFAOYSA-N

General description

A cell-permeable powerful DNA topoisomerase I inhibitor. Reduces DNA synthesis in vitro in murine hepatoma cells. Has selective inhibitory effect on the phosphorylation of histone H1 and H3, but less effect on other histones.
A cell-permeable, powerful DNA topoisomerase I inhibitor. Reduces DNA synthesis in vitro in murine hepatoma cells. Inhibits the proliferation of hepatoma HepG2, Bcl-7402 and Bcl-7404 cells in a dose-dependent manner. Has selective inhibitory effect on the phosphorylation of histones H1 and H3, but is less effective on other histones.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
DNA topoisomerase I
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Zhang, X.W., et al. 1999. Anticancer Drugs.10, 569.
Ling, Y.H., et al. 1993. Anticancer Res. 13, 1613.
Wani, M.C., and Wall, M.E. 1969. J. Org.Chem.34, 1364.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Venugopal R Bovilla et al.
Biomedicines, 9(9) (2021-09-29)
Nuclear factor erythroid-2 related factor-2 (Nrf2) is an oxidative stress-response transcriptional activator that promotes carcinogenesis through metabolic reprogramming, tumor promoting inflammation, and therapeutic resistance. However, the extension of Nrf2 expression and its involvement in regulation of breast cancer (BC) responses
Mengfan Tang et al.
Nucleic acids research, 49(13), 7476-7491 (2021-07-02)
Poly (ADP-ribose) polymerase inhibitor (PARPi)-based therapies initially reduce tumor burden but eventually lead to acquired resistance in cancer patients with BRCA1 or BRCA2 mutation. To understand the potential PARPi resistance mechanisms, we performed whole-genome CRISPR screens to discover genetic alterations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service