Skip to Content
Merck
All Photos(1)

Key Documents

329600

Sigma-Aldrich

Erastin

≥98% (HPLC), solid, ferroptosis inducer, Calbiochem®

Synonym(s):

Erastin, 2-(1-(4-(2-(4-Chlorophenoxy)acetyl)-piperazin-1-yl)-ethyl)-3-(2-ethoxyphenyl)-3H-quinazolin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H31ClN4O4
CAS Number:
Molecular Weight:
547.04
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Erastin, A cell-permeable piperazinyl-quinazolinone compound that exhibits oncogene-selective lethality.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

Clc1ccc(cc1)OCC(=O)N2CCN(CC2)C(C)c3[n]([c](c5c(n3)cccc5)=O)c4c(cccc4)OCC

InChI

1S/C30H31ClN4O4/c1-3-38-27-11-7-6-10-26(27)35-29(32-25-9-5-4-8-24(25)30(35)37)21(2)33-16-18-34(19-17-33)28(36)20-39-23-14-12-22(31)13-15-23/h4-15,21H,3,16-20H2,1-2H3

InChI key

BKQFRNYHFIQEKN-UHFFFAOYSA-N

General description

A cell-permeable piperazinyl-quinazolinone compound that exhibits oncogene-selective lethality. Shown to bind mitochondrial voltage-dependent anion channels (VDAC) and alter its gating; rapidly induce an oxidative, non-apoptotic cell death in several human tumors harboring activating mutations in the RAS-RAF-MEK signaling (EC50 = 6.0 µM in BJ-TERT/LT/ST/RASV12 cells).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Yagoda, N., et al. 2007. Nature447, 864.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhuhui Yuan et al.
Oxidative medicine and cellular longevity, 2022, 3403009-3403009 (2022-10-21)
Ionizing radiation-derived oxidative stress and ferroptosis are one of the most important biological effects on destroying the liver tumor, whereas radioresistance of liver tumor remains a leading cause of radiotherapy (RT) failure mainly because of the protective antiferroptosis, in which
Natalie K Horvat et al.
ACS nano, 18(17), 11025-11041 (2024-04-17)
ALK-positive NSCLC patients demonstrate initial responses to ALK tyrosine kinase inhibitor (TKI) treatments, but eventually develop resistance, causing rapid tumor relapse and poor survival rates. Growing evidence suggests that the combination of drug and immune therapies greatly improves patient survival;
Zhenyi Su et al.
Nature cancer, 4(4), 564-581 (2023-03-28)
Although the gain of function (GOF) of p53 mutants is well recognized, it remains unclear whether different p53 mutants share the same cofactors to induce GOFs. In a proteomic screen, we identified BACH1 as a cellular factor that recognizes the
DHODH inhibitors sensitize to ferroptosis by FSP1 inhibition.
Eikan Mishima et al.
Nature, 619(7968), E9-E18 (2023-07-06)
Jiashuo Zheng et al.
Cell death & disease, 12(7), 698-698 (2021-07-15)
Sorafenib, a protein kinase inhibitor approved for the treatment of hepatocellular carcinoma and advanced renal cell carcinoma, has been repeatedly reported to induce ferroptosis by possibly involving inhibition of the cystine/glutamate antiporter, known as system xc-. Using a combination of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service