Skip to Content
Merck
All Photos(1)

Key Documents

236600

Sigma-Aldrich

C-Reactive Protein, Human Ascites

C - reactive protein, Human Ascites, CAS 9007-41-4, is a major acute-phase plasma protein that composed of five identical non-covalently linked subunits.

Synonym(s):

C-Reactive Protein, Human Ascites, CRP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14, 2603

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: soluble

shipped in

wet ice

storage temp.

2-8°C

General description

M.W. 23,000 (monomer); 115,000 (pentamer).
Native C-reactive protein from human ascites. CRP is a major acute-phase plasma protein that is dramatically elevated in patients with acute-phase conditions. Composed of five identical noncovalently linked subunits. Exhibits Ca2+-dependent binding to a variety of substrates. Important in diagnosis of rheumatoid arthritis. Suitable for immunological studies. In purified states and in plasma the monomeric form (M.W. ~21 kDa) aggregates as a cyclic pentamer.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 20 mM Tris, 2 mM CaCl₂, 0.1% NaN₃, pH 7.5.

Preparation Note

Prepared from ascites that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Analysis Note

Single band by SDS-PAGE

Other Notes

Christner, R.B., and Mortensen, R.F. 1994. J. Biol. Chem. 269, 9760.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katrine Pilely et al.
Frontiers in immunology, 8, 1040-1040 (2017-09-14)
Inflammation is a part of the initial process leading to atherosclerosis and cholesterol crystals (CC), found in atherosclerotic plaques, which are known to induce complement activation. The pentraxins C-reactive protein (CRP), long pentraxin 3 (PTX3), and serum amyloid P component

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service