Skip to Content
Merck
All Photos(1)

Key Documents

234610

Sigma-Aldrich

Congo Red, High Purity

An amyloidophylic dye that specifically stains stacked β sheet aggregates.

Synonym(s):

Congo Red, High Purity, 3,3ʹ-[(1,1ʹ-Biphenyl)-4,4ʹ-diylbis(azo)]bis(4-amino-1-naphthalenesulfonic Acid) 2Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H22N6O6S2 · 2Na
CAS Number:
Molecular Weight:
696.66
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.25

Quality Level

description

Merck USA index - 14, 2498

Assay

≥75% dye basis

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

rust

solubility

water: 10 mg/mL
ethanol: soluble

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Na+].[Na+].[S](=O)(=O)([O-])c1c2c(c(c(c1)N=Nc3ccc(cc3)c4ccc(cc4)N=Nc5c(c6c(c(c5)[S](=O)(=O)[O-])cccc6)N)N)cccc2

InChI

1S/C32H24N6O6S2.2Na/c33-31-25-7-3-1-5-23(25)29(45(39,40)41)17-27(31)37-35-21-13-9-19(10-14-21)20-11-15-22(16-12-20)36-38-28-18-30(46(42,43)44)24-6-2-4-8-26(24)32(28)34;;/h1-18H,33-34H2,(H,39,40,41)(H,42,43,44);;/q;2*+1/p-2

InChI key

IQFVPQOLBLOTPF-UHFFFAOYSA-L

General description

An amyloidophylic dye that specifically stains stacked β sheet aggre-gates. Useful for staining and visual detection of amyloid deposits in nerve tissue. Binds to amyloid fibrils but not to non-polymerized amyloid peptide. Inhibits the accumulation of the scrapie-associated, protease-resistant isoform of protein PrP without affecting the normal isoform possibly through overstabilization of the conformation. It is also shown to inhibit Pot1 Binding to Telomeric DNA.
An amyloidophylic dye that specifically stains stacked β sheet aggregates. Useful for staining and visual detection of amyloid deposits. Does not bind to non-polymerized amyloid peptide. Inhibits the accumulation of the scrapie-associated, protease-resistant isoform of protein PrP without affecting the metabolism of the normal isoform. It is also shown to inhibit Pot1 Binding to Telomeric DNA.

Biochem/physiol Actions

Cell permeable: no
Primary Target
An amyloidophylic dye that specifically stains stacked β sheet aggregates
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at room temperature (20°C).

Other Notes

Altschuler, S.E., et al. 2012. Biochemistrin press.
Khurana, R., et al. 2001. J. Biol. Chem.276, 22715.
Fezoui, Y., et al. 2000. Amyloid7, 166.
Fezoui, Y., et al. 2000. Nat. Struct. Biol.7, 1095.
Kramport, M., et al. 2000. Chem. Biol.7, 855.
Kuner, P., et al. 2000. J. Biol. Chem.275, 1673.
Caspi, S., et al. 1998. J. Biol. Chem.273, 3484.
Caughey, B., and Race, A.E. 1992. J. Neurochem.59, 768.
Taylor, D.L., et al. 1974. J. Histochem. Cytochem.22, 1105.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service