Skip to Content
Merck
All Photos(1)

Key Documents

208745

Sigma-Aldrich

Calpain Inhibitor VI

The Calpain Inhibitor VI, also referenced under CAS 190274-53-4, controls the biological activity of Calpain. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Calpain Inhibitor VI, N-(4-Fluorophenylsulfonyl)-L-valyl-L-leucinal, SJA6017

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25FN2O4S
CAS Number:
Molecular Weight:
372.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Fc1ccc(cc1)[S](=O)(=O)NC(C(C)C)C(=O)NC(CC(C)C)C=O

InChI

1S/C17H25FN2O4S/c1-11(2)9-14(10-21)19-17(22)16(12(3)4)20-25(23,24)15-7-5-13(18)6-8-15/h5-8,10-12,14,16,20H,9H2,1-4H3,(H,19,22)

InChI key

WSJWUIDLGZAXID-UHFFFAOYSA-N

General description

A cell-permeable peptide aldehyde that acts as a potent and reversible inhibitor of calpain (IC50 = 7.5 nM for µ-calpain and 78 nM for m-calpain). Also potently inhibits cathepsin B (IC50 = 15 nM) and cathepsin L (IC50 = 1.6 nM). Reduces βFGF-induced angiogenesis in rat cornea and prevents selenite cataract formation in rats. Reduces A23187-induced nuclear opacity and proteolysis of crystallins and α-spectrin in cultured rat lenses.
A potent, cell-permeable, reversible inhibitor of calpain (IC50 = 7.5 nM for µ-calpain and 78 nM for m-calpain). Also potently inhibits cathepsin B (IC50 = 15 nM) and L (IC50 = 1.6 nM). Reduces bFGF-induced angiogenesis in rat cornea and prevents selenite cataract formation. Reduces A23187-induced nuclear opacity and proteolysis of crystallins and α-spectrin in cultured lenses.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
µ-calpain
Product does not compete with ATP.
Reversible: yes
Target IC50: 7.5 nM for µ-calpain and 78 nM for m-calpain; 15 nM and 1.6 nM against cathepsin B and L, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

4-Fluorophenylsulfonyl-Val-Leu-CHO

Reconstitution

Following reconstitution, aliquot and freeze at -20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Inoue, J., et al. 2003. J. Med. Chem.46, 868.
Tamada, Y., et al. 2001. Curr. Eye Res.22, 280.
Mathur, P., et al. 2000. Curr. Eye Res.21, 926.
Nath, R., et al. 2000. Biochem. Biophys. Res. Commun.274, 16.
Tamada, Y., et al. 2000. J. Ocul. Pharmacol. Ther.16, 271.
Fukiage, C., et al. 1998. Curr. Eye Res.17, 623.
Fukiage, C., et al. 1997. Biochim. Biophys. Acta 1361, 304.

Legal Information

Sold under license of U.S. Patent 6,551,999
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service