Skip to Content
Merck
All Photos(1)

Key Documents

178479

Sigma-Aldrich

Anti-Apolipoprotein E Goat pAb

liquid, Calbiochem®

Synonym(s):

Anti-ApoE antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, rat, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Apolipoprotein E, goat polyclonal, recognizes human apolipoprotein E. Does not cross-react with other apolipoproteins. It is validated for ELISA, immunoprecipitation, and free-floating sections.
Goat polyclonal antibody supplied as serum that has been defibrinated, delipidized and dialyzed against a Tris-HCl buffer. Recognizes the apolipoprotein E protein.
Recognizes human apolipoprotein E. Does not cross-react with other apolipoproteins.
  • Antibody Target Gene Symbol: APOE
  • Target Synonym: AD2, AI255918, APOE2, APOE4, APOEA, APOLIPOPROTEIN E, LDLCQ5, LPG, MGC1571
  • Entrez Gene Name: apolipoprotein E
  • Hu Entrez ID: 348 (Related Antibodies: NE1004)
  • Mu Entrez ID: 11816
  • Rat Entrez ID: 25728
  • Immunogen

    Human
    purified recombinant human apolipoprotein E

    Application

    ELISA (1:8000)

    Immunoprecipitation (see comments)

    Free Floating Sections (see application references)

    Packaging

    Please refer to vial label for lot-specific concentration.

    Warning

    Toxicity: Standard Handling (A)

    Physical form

    In 500 mM NaCl, 50 mM Tris-HCl, pH 7.5.

    Reconstitution

    Following initial thaw, aliquot and freeze (-20°C).

    Other Notes

    Monospecific as determined by immunoelectrophoresis (IEP) against twice concentrated pooled human serum. This antibody has also been reported to work with immunoprecipitation. Variables associated with assay conditions will dictate the proper working dilution.

    Legal Information

    CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

    Not finding the right product?  

    Try our Product Selector Tool.

    Storage Class Code

    10 - Combustible liquids

    WGK

    WGK 1

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Emilie L Castranio et al.
    Neurobiology of disease, 105, 1-14 (2017-05-16)
    Traumatic brain injury (TBI) is strongly linked to an increased risk of developing dementia, including chronic traumatic encephalopathy and possibly Alzheimer's disease (AD). APOEε4 allele of human Apolipoprotein E (APOE) gene is the major genetic risk factor for late onset
    Jin-Dong Ding et al.
    Proceedings of the National Academy of Sciences of the United States of America, 108(28), E279-E287 (2011-06-22)
    Age-related macular degeneration (AMD) is a leading cause of visual dysfunction worldwide. Amyloid β (Aβ) peptides, Aβ1-40 (Aβ40) and Aβ1-42 (Aβ42), have been implicated previously in the AMD disease process. Consistent with a pathogenic role for Aβ, we show here
    Suzanne E Wahrle et al.
    The Journal of clinical investigation, 118(2), 671-682 (2008-01-19)
    APOE genotype is a major genetic risk factor for late-onset Alzheimer disease (AD). ABCA1, a member of the ATP-binding cassette family of active transporters, lipidates apoE in the CNS. Abca1(-/-) mice have decreased lipid associated with apoE and increased amyloid
    Nicholas F Fitz et al.
    Journal of Alzheimer's disease : JAD, 41(2), 535-549 (2014-03-20)
    Passive amyloid-β (Aβ) vaccination has shown significant effects on amyloid pathology in pre-depositing amyloid-β protein precursor (AβPP) mice but the results in older mice are inconsistent. A therapeutic effect of LXR and RXR agonists consisting of improved memory deficits and
    Elizabeth S Chan et al.
    Scientific reports, 6, 26119-26119 (2016-05-18)
    The apolipoprotein E4 (ApoE4) is the strongest genetic risk factor for Alzheimer's disease (AD). The AD brain was shown to be insulin resistant at end stage, but the interplay between insulin signaling, ApoE4 and Aβ across time, and their involvement

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service