17-375
EZ-Zyme™ Chromatin Prep Kit
Contains proprietary reagents optimized for the enzymatic shearing of chromatin from mammalian cells at higher resolution than sonication for use in chromatin immunoprecipitation (ChIP) assays.
Synonym(s):
Chromatin Isolation Kit
About This Item
Recommended Products
Quality Level
manufacturer/tradename
EZ-Zyme™
Upstate®
application(s)
sample preparation
shipped in
dry ice
Related Categories
General description
When performing ChIP, chromatin from cells and tissues needs to be fragmented so that it becomes soluble and resolution can be achieved in detecting protein-DNA interaction at specific loci. Sonication is a common method for producing sheared chromatin. Although this method is very efficient in releasing chromatin into the solution, it has a number of limitations. For example, foaming and overheating associated with sonication can result in protein denaturation or incomplete chromatin fragmentation. Also, sonication conditions can be physically harsh and can negatively impact immunoprecipitation efficiency. These and other limitations in the procedure can make sample sonication difficult to optimize and perform reproducibly.
The EZ-Zyme Chromatin Prep kit allows ChIP analysis at nucleosome resolution by performing complete or partial digestions with a proprietary enzymatic cocktail to obtain chromatin fragments of on average one to a few nucleosomes in length. Compared to sonication, this procedure not only allows mapping the protein-DNA association at a higher resolution, but also renders the subsequent immunoprecipitation more efficient due to its mild digestion condition. In addition, enzymatic shearing is compatible with non-crosslinked ChIP (native ChIP), which allows analysis of histone modifications not only on cultured cells, but also on freshly dissected and frozen tissues.
Use:
The EZ-Zyme kit contains reagents optimized for generating enzymatically processed chromatin from mammalian cells for use in chromatin immunoprecipitation. While the kit contains a full complement of buffers required for chromatin shearing, careful attention must be paid to the details of the instructions. Follow all the instructions carefully, especially with regard to incubation times and temperatures.
Packaging
Components
0.5M EDTA, Catalog # 20-158. One vial containing 250 μL of 0.5M EDTA, pH 8.0.
5M NaCl, Catalog # 20-159. One vial containing 500 μL of 5M NaCl.
1M Tris-HCl, pH 6.5, Catalog # 20-160. One vial containing 500 μL of 1M Tris-HCl, pH 6.5.
10X Glycine, Catalog # 20-282. One vial containing 11 mL of 1.25M Glycine.
10X PBS, Catalog # 20-281. One vial containing 24 mL of 10X PBS.
EZ-Zyme Lysis Buffer, Catalog # 20-312. One vial containing 2.0 mL of EZ-Zyme Lysis Buffer.
EZ-Zyme Digestion Buffer, Catalog # 20-313. One vial containing 1.65 mL of EZ-Zyme Digestion Buffer.
EZ-Zyme Stop Buffer, Catalog # 20-315. One vial containing 1.65 mL of EZ-Zyme Stop Buffer
Store at -20°C:
Protease Inhibitor Cocktail II, Catalog # 20-283. One vial containing 110 μL of Protease Inhibitor Cocktail II in DMSO.
RNase A, Catalog # 20-297. One vial containing 600 μg of RNase A in 60 μL sterile water.
Proteinase K, Catalog # 20-298. One vial containing 600 μg of Proteinase K in 60 μL of 50mM Tris-HCl, pH 8.0, 10mM CaCl2.
EZ-Zyme Enzymatic Cocktail, Catalog # 20-314. One vial containing 76U of Enzymatic Cocktail in 38 μL of a solution containing 50% glycerol.
Storage and Stability
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service