Skip to Content
Merck
All Photos(1)

Key Documents

14-751-M

Sigma-Aldrich

Abl (Q252H) Protein, active, 10 µg

Active, N-terminal 6His-tagged recombinant human Abl, residues 27-end containing the Q252H mutation. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

100
300

recombinant

expressed in baculovirus infected Sf21 cells

specific activity

1089 U/mg

mol wt

Mw 121 kDa

manufacturer/tradename

Upstate®

concentration

51% (Coomassie staining, SDS-PAGE)

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

N-terminal 6His-tagged recombinant human Abl, residues 27-end containing the Q252H mutation. The Abl tyrosine kinase inhibitor STI571 is an effective therapy for stable-phase chronic myeloid leukemia patients. Many patients responding to STI571 later relapse due to a reactivation of Bcr-Abl activity. In certain cases this appears to correlate with the presence of the Q252H mutation, which confers resistance to drug binding (Shah et al., (2002), Cancer Cell 2, 117-125).
Product Source: Expressed by baculovirus in Sf21 insect cells.

Application

Research Category
Metabolism
Research Sub Category
Cardiovascular Disease

Metabolic Disorders

Biochem/physiol Actions

Protein Target: Abl
Target Sub-Family: TK

Quality

Routinely evaluated by phosphorylation of 50 μM Abltide. Purity was assessed by SDS-PAGE and Coomassie blue staining using 3 μg of active Abl (Q252H).

Unit Definition

Specific Activity: one unit of Abl (Q252H) activity is defined as 1 nmol phosphate incorporated into 50 μM Abltide (EAIYAAPFAKKK) per minute at 30°C with a final ATP concentration of 100 μM.

Physical form

10 μg of enzyme in 100 μL of 50 mM Tris/HCl pH7.5, 150 mM NaCl, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij 35. Frozen solution.
Ni2+/NTA-agarose

Storage and Stability

Store -70°C. For maximum recovery of product, centrifuge original vial prior to removing the cap.
Rapidly thaw the vial under cold water and immediately place on ice. Aliquot unused material into pre-chilled microcentrifuge tubes and immediately snap-freeze the vials in liquid nitrogen prior to re-storage at -70°C.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service