1468510006
Tryptic Soy Broth Ready-to-use Bottles
Tween® 0.1 %, bottle capacity 1 L, bottle filling volume 1 L, closure type, blue flip cap with septum, box of 6 bottles
Synonym(s):
Casein Soyabean Digest Broth, SCDB, TSB, TSB with 0.1% Tween®80, Tryptone Soy Broth
About This Item
Recommended Products
form
liquid
Quality Level
feature
closure type blue flip cap with septum
composition
Tween®, 0.1%
packaging
box of 6 bottles
bottle capacity
1 L
bottle filling volume
1 L
color
light yellow
pH
7.3±0.2 (25 °C)
application(s)
food and beverages
pharmaceutical
sterility testing
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | 1.46802 | 1.46627 | 1.46283 |
---|---|---|---|
food and beverages | food and beverages | food and beverages | cosmetics |
liquid | liquid | liquid | liquid (ready-to-use) |
box of 6 bottles | box of 6 bottles, bottle of | box of 10 bottles | box of 20 or 100 tubes, tube of |
closure type blue flip cap with septum | closure type blue screw cap with septum, ready-to-use | closure type blue screw cap with 3 loci, ready-to-use | closure type blue screw cap with 2 loci, ready-to-use |
2-25°C | 2-25°C | 2-25°C | 2-25°C |
General description
Upon incubation, the enumerated microflora can be plated, isolated, and identified.
Application
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service