Skip to Content
Merck
All Photos(1)

Key Documents

1.46465

Millipore

Yeast Glucose Chloramphenicol Agar

Determination of the colony count of yeasts and molds in dairy and other food products

Synonym(s):

YG Agar with Chloramphenicol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106227
NACRES:
NA.74

Pricing and availability is not currently available.

Quality Level

feature

ready-to-use

packaging

pkg of (200 ml in 250 ml bottle with blue screw cap and 3 loci (6 bottles per box))

pH

6.6 ( in H2O)

application(s)

food and beverages
microbiology

storage temp.

2-25°C

suitability

molds
yeasts

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
1.462371.009181.13116
application(s)

food and beverages
microbiology

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

application(s)

cosmetics
food and beverages
pharmaceutical

application(s)

food and beverages
microbiology

Quality Level

300

Quality Level

300

Quality Level

200

Quality Level

200

storage temp.

2-25°C

storage temp.

2-8°C

storage temp.

15-25°C

storage temp.

15-25°C

pH

6.6 ( in H2O)

pH

5.6 ( in H2O)

pH

5.6±0.2 (25 °C)

pH

-

feature

ready-to-use

feature

ready-to-use

feature

-

feature

-

General description

Yeast Glucose Chloramphenicol (YGC) Agar is suitable for the determination of the colony count of yeasts and molds in food products and Beverage. Yeast extract provides the necessary carbon and nitrogen compounds, vitamins and trace elements for the growth of microorganisms. Glucose also serves as a carbon and energy source, the high concentration at the same time promotes the fungal growth and inhibits bacterial growth. Chloramphenicol inhibits the accompanying bacterial flora. The neutral pH facilitates the growth of stressed yeast and molds.

Application

Yeast Glucose Chloramphenicol (YGC) Agar is suitable for the determination of the colony count of yeasts and molds in food products and Beverage

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Need A Sample COA?

    This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service