Skip to Content
Merck
All Photos(1)

Key Documents

1.46425

Millipore

Orange Serum Agar

Detection of bacterial count and confirmation of the presence of acid-tolerant microorganisms in food packaging materials and beverages

Synonym(s):

Orange Serum Medium, OSA

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41106228
NACRES:
NA.74

form

solid (ready-to-use)

feature

ready-to-use

packaging

pkg of (200 ml in 250 ml bottle with blue screw cap and 3 loci (6 bottles per box))

pH

5.5 ( in H2O)

application(s)

food and beverages
microbiology

storage temp.

2-25°C

General description

Orange Serum Agar is used for the isolation, cultivation and bacterial count of acid-tolerant microorganisms that cause deterioration of fruit juices and fruit-juice concentrates, particularly those of citrus fruits. It is generally used in fruit-juice production and to confirm the presence of acid-tolerant microorganisms in food packaging materials. Casein peptone serves the growth requirements such as carbon and nitrogen compounds, yeast extract provides B-complex vitamins and trace elements. Glucose is also a source of carbon and energy. In combination with the low pH value of the medium, the additional acid formation from glucose favors the growth of yeasts, molds, Lactobacillus sp. and Leuconostoc sp. These spoilage agents are particularly stimulated by the addition of orange extract (orange serum).

Application

Orange Serum Agar is used for the isolation, cultivation and bacterial count of acid-tolerant microorganisms in fruit juices and fruit-juice concentrates, particularly those of citrus fruits and food packaging materials.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service