Skip to Content
Merck
All Photos(1)

Key Documents

13-104

Sigma-Aldrich

MBP (myelin basic protein), bovine, purified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

biological source

bovine

Quality Level

form

liquid

shelf life

2 yr

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

solubility

water: soluble

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

bovine ... MBP(618684)

General description

Myelin basic protein (MBP) belongs to the intrinsically disordered protein family. It is localized in the insulating layer of nerve cells in the brain and spinal cord. It is an abundant protein found in the central nervous system (CNS).
Research area: Cell Signaling
MBP is an alkaline protein with a pI of 11.6, constituting approximately 30% of the total protein content of the myelin in the central nervous system.

Application

MBP (myelin basic protein), bovine, purified has been used in western blotting and as a standard in enzyme-linked immunosorbent assay (ELISA).

Biochem/physiol Actions

MBP can interact with various polyanionic proteins such as actin, tubulin, Ca2+-calmodulin, and clathrin, as well as negatively charged lipids. Upon binding to these proteins, MBP undergoes structural changes. It is believed to function as a membrane actin-binding protein, potentially enabling its involvement in relaying extracellular signals to the cytoskeleton in oligodendrocytes and tight junctions in myelin. External signals received by myelin or cultured oligodendrocytes lead to alterations in the phosphorylation of MBP, indicating its participation in signaling. The peptide MBP4-14 is specific for protein kinase C and is resistant to phosphatase activity. Moreover, MBP is extensively utilized in the "in gel" kinase assay, making it a vital biochemical for studying protein kinase activities.
Myelin basic protein (MBP), known as the ‘executive molecule of myelin′, helps in the formation of central nervous system (CNS) myelin. Elevated levels of MBP in the cerebrospinal fluid (CSF) are detected in acute neurological disorders like exacerbating multiple sclerosis, and traumatic brain injury. It is also observed at increased levels during chronic progressive disorders including dementia, tumor cerebri, and metabolic disorders. 

Quality

routinely evaluated by successfully phosphorylating MAP Kinase 2/Erk2, active (Catalog # 14-173)

Physical form

10mM MOPS, pH 7.0, with 0.05% sodium azide

Storage and Stability

2 years at -20°C

Other Notes

This product is derived from bovine source. Export of this product to certain countries may be restricted. Please contact Customer Service or your local distributor to inquire about product availability and export options.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A C Maroney et al.
The Journal of biological chemistry, 276(27), 25302-25308 (2001-04-28)
CEP-1347 (KT7515) promotes neuronal survival at dosages that inhibit activation of the c-Jun amino-terminal kinases (JNKs) in primary embryonic cultures and differentiated PC12 cells after trophic withdrawal and in mice treated with 1-methyl-4-phenyl tetrahydropyridine. In an effort to identify molecular
Dajun Sang et al.
eLife, 8 (2019-08-14)
Protein kinases are crucial to coordinate cellular decisions and therefore their activities are strictly regulated. Previously we used ancestral reconstruction to determine how CMGC group kinase specificity evolved (Howard et al., 2014). In the present study, we reconstructed ancestral kinases
Purification of myelin basic protein from bovine brain
Chevalier, Dominique and Allen, Bruce G
Protein Expression and Purification, 18(2), 229-234 (2000)
Tuoping Luo et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(8), 2860-2865 (2012-02-11)
Approximately 30% of human cancers harbor oncogenic gain-of-function mutations in KRAS. Despite interest in KRAS as a therapeutic target, direct blockade of KRAS function with small molecules has yet to be demonstrated. Based on experiments that lower mRNA levels of
Myelin basic protein: a multifunctional protein
Boggs JM
Cellular and Molecular Life Sciences (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service