Skip to Content
Merck
All Photos(1)

Key Documents

1.15368

Supelco

TISAB solution

Total ionic strength ajustment buffer, for fluoride determination

Synonym(s):

Total Ionic Strength Adjustment Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401
NACRES:
NA.21

product name

TISAB solution, for fluoride determination

Quality Level

form

liquid

technique(s)

titration: suitable

pH

5.5 (20 °C in H2O)

density

1.07 g/cm3 at 20 °C

storage temp.

15-25°C

Application


  • Bupropion HCl research solution: Utilized in neuropsychiatric drug toxicity studies to assess the mitigating effects of lipid emulsions on drug-induced adverse effects (Hwang and Sohn, 2024).

  • Bupropion biochemistry application: Investigated for its anti-inflammatory effects in relation to stress and depression, providing insights into its biochemical impact on low-grade chronic inflammation (Mehdi et al., 2023).

  • Bupropion hydrochloride pharmacological research: Examined for potential therapeutic effects on anxiety and depression related to nicotine withdrawal, focusing on its influence on inflammatory, oxidative, and serotoninergic profiles (Pahlavani et al., 2023).



Analysis Note

Assay (NaCl): 58.0 - 60.0 g/l
Assay (Titriplex IV): 3.6 - 4.0 g/l
pH-value (20 °C): 5.0 - 6.0
Fluoride (F): ≤ 0.1 µg/ml

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service