1.12230
Lithium nitrate
98+
Synonym(s):
Lithium nitrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
LiNO3
CAS Number:
Molecular Weight:
68.95
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-218-9
NACRES:
NA.22
solubility:
522 g/L
Recommended Products
Quality Level
pH
7-9 (20 °C, 50 g/L in H2O)
mp
255 °C
solubility
522 g/L
density
2.36 g/cm3 at 20 °C
bulk density
910 kg/m3
storage temp.
2-30°C
SMILES string
[Li+].[N+](=O)([O-])[O-]
InChI
1S/Li.NO3/c;2-1(3)4/q+1;-1
InChI key
IIPYXGDZVMZOAP-UHFFFAOYSA-N
Application
- The critical role of lithium nitrate in the gas evolution of lithium–sulfur batteries: Lithium nitrate is used as a stabilizing additive in lithium-sulfur batteries, impacting gas evolution related to the lithium metal anode (A Jozwiuk et al., 2016).
- Thermal characterisation of binary sodium/lithium nitrate salts for latent heat storage at medium temperatures: Investigates the thermal performance of binary sodium/lithium nitrate salts, crucial for latent heat storage applications (D Zhou, P Eames, 2016).
- The synergetic effect of lithium polysulfide and lithium nitrate to prevent lithium dendrite growth: Discusses the combined use of lithium polysulfide and lithium nitrate as additives to mitigate lithium dendrite growth in batteries (W Li et al., 2015).
- Nitrate additives for lithium batteries: Mechanisms, applications, and prospects: Reviews the use of nitrate additives like lithium nitrate in lithium batteries, focusing on mechanisms and performance (X Li et al., 2021).
- Stable all-solid-state lithium metal batteries with Li3N-LiF-enriched interface induced by lithium nitrate addition: Studies the effect of lithium nitrate on enhancing the stability of lithium metal batteries through interface modifications (Z Zhang et al., 2021).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service