Skip to Content
Merck
All Photos(1)

Key Documents

1.10897

Sigma-Aldrich

N,N′-Methylenediacrylamide

for electrophoresis

Synonym(s):

N,N′-Methylenediacrylamide, BIS, Bisacrylamide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CHCONH)2CH2
CAS Number:
Molecular Weight:
154.17
MDL number:
UNSPSC Code:
41105305
EC Index Number:
203-750-9
NACRES:
NC.01
technique(s):
electrophoresis: suitable
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (saponification, acidimetric)

form

crystalline powder

potency

390 mg/kg LD50, oral (Rat)

technique(s)

electrophoresis: suitable

pH

≥5.0 (20 °C, 25 g/L in H2O)

mp

185 °C (decomposition)

density

1.235 g/cm3 at 30 °C

bulk density

200 kg/m3

storage temp.

2-8°C

SMILES string

N(CNC(=O)C=C)C(=O)C=C

InChI

1S/C7H10N2O2/c1-3-6(10)8-5-9-7(11)4-2/h3-4H,1-2,5H2,(H,8,10)(H,9,11)

InChI key

ZIUHHBKFKCYYJD-UHFFFAOYSA-N

Analysis Note

Appearance (colour): white
Appearance (description): crystalline powder
Spec. Absorptivity A 1%/1cm (290 nm; 1 %; 1 cm; water): ≤ 0.200
pH-value (2.5 %, water): ≥ 5.0
Assay (by acidimetry following saponification): ≥ 99.0 %
Identity (IR-spectrum): passes test
Heavy metals (as Pb): ≤ 0.001 %
Free acid (as acrylic acid): ≤ 0.02 %
Water: ≤ 0.5 %

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 1B - Muta. 1B - Repr. 2 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service