Skip to Content
Merck
All Photos(1)

Key Documents

1.09947

Supelco

Silicon standard

1000 mg Si, (SiCl₄ in 14% NaOH) Titrisol®

Synonym(s):

Silicon Reference Standard for ICP/AAS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

Titrisol®

form

concentrate

concentration

1000 mg Si in 14% NaOH

technique(s)

AAS: suitable
ICP: suitable

pH

14 (20 °C in H2O)

density

1.15 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

Features and Benefits

  • TitrisolTitrisol® offers a wide range of concentrated single-element standards.
  • Dilute the concentrated solution to achieve the desired standard concentration.
  • Directly traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST).
  • Supplied in easy-to-handle plastic ampoules.
  • Accompanied with a batch-specific certificate of analysis (COA) containing information on the NIST batch and minimum shelf life.
  • Provides reliable and reproducible results.
  • Analysis Note

    content (Si): 1000 mg / Ampoule
    Determination method: ICP-OES
    (traceable to NIST - SRM 3150)
    Accuracy of the method: +/- 5 mg

    Legal Information

    Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

    Pictograms

    Corrosion

    Signal Word

    Danger

    Hazard Statements

    Hazard Classifications

    Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

    Supplementary Hazards

    Storage Class Code

    8B - Non-combustible, corrosive hazardous materials

    WGK

    WGK 1

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service