1.09630
2-Butanol
for analysis EMSURE®
Synonym(s):
2-Butanol, sec-Butyl alcohol
About This Item
Recommended Products
vapor pressure
16.5 hPa ( 20 °C)
Quality Level
product line
EMSURE®
Assay
≥99% (GC)
form
liquid
autoignition temp.
390 °C
potency
6480 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)
expl. lim.
1.4-9.8 % (v/v)
impurities
≤0.00005% Al (Aluminium)
≤0.0002 meq/g Alkalinity
≤0.0005 meq/g Acidity
≤0.1% Ethylmethylketone (GC)
≤0.1% tert-butanol (GC)
≤0.2% 2-Propanol (GC)
≤0.2% Dibutyl ether (GC)
evapn. residue
≤0.001%
color
APHA: ≤10
pH
7 (20 °C)
bp
99-102 °C/1013 hPa
mp
-115 °C
transition temp
flash point 24 °C
solubility
125 g/L
density
0.81 g/cm3 at 20 °C
cation traces
Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%
storage temp.
2-30°C
SMILES string
OC(CC)C
InChI
1S/C4H10O/c1-3-4(2)5/h4-5H,3H2,1-2H3
InChI key
BTANRVKWQNVYAZ-UHFFFAOYSA-N
General description
Analysis Note
Identity (IR): conforms
Colour: ≤ 10 Hazen
Acidity: ≤ 0.0005 meq/g
Alkalinity: ≤ 0.0002 meq/g
tert-butanol (GC): ≤ 0.1 %
Dibutyl ether (GC): ≤ 0.2 %
Ethylmethylketone (GC): ≤ 0.1 %
2-Propanol (GC): ≤ 0.2 %
Al (Aluminium): ≤ 0.00005 %
B (Boron): ≤ 0.000002 %
Ba (Barium): ≤ 0.00001 %
Ca (Calcium): ≤ 0.00005 %
Cd (Cadmium): ≤ 0.000005 %
Co (Cobalt): ≤ 0.000002 %
Cr (Chromium): ≤ 0.000002 %
Cu (Copper): ≤ 0.000002 %
Fe (Iron): ≤ 0.00001 %
Mg (Magnesium): ≤ 0.00001 %
Mn (Manganese): ≤ 0.000002 %
Ni (Nickel): ≤ 0.000002 %
Pb (Lead): ≤ 0.00001 %
Sn (Tin): ≤ 0.00001 %
Zn (Zinc): ≤ 0.00001 %
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.2 %
Other Notes
Legal Information
accessory
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
80.6 °F - closed cup
Flash Point(C)
27 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service