Skip to Content
Merck
All Photos(1)

Documents

1.09293

Millipore

KOVACS′ indole reagent

for detecting microbial indole in the identification of indole-positive and indole-negative microorganisms, suitable for microbiology

Synonym(s):

4-(Dimethylamino)benzaldehyde solution, Indole reagent, Kovac′s reagent, Kovac′s reagent for indoles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.21

Quality Level

form

liquid

packaging

pkg of 100 mL

technique(s)

microbe id | specific enzyme detection: suitable

color

yellow

pH

<1 (20 °C in H2O)

transition temp

flash point 36 °C

density

0.92 g/cm3 at 20 °C

application(s)

microbiology

storage temp.

2-8°C

suitability

Escherichia coli
enterovirulent E. coli (EEC) spp.

General description

Kovac′s reagent is a biochemical reagent composed of n-Butanol, hydrochloric acid, and 4-dimethylaminobenzaldehyde. It is used for the detection of indole producing microorganisms. Tryptophan is abundant in trypticalle digested peptone. Some microorganisms can cleave tryptophan to give pyruvic acid, ammonia, and indole. Indole then reacts with 4-dimethylaminobenzaldehyde to form a dark red dye. As tryptophan also gives a color reaction with 4 dimethylaminobenzaldehyde, it must be separated from the indole. This is achieved by selectively extracting indole with butanol.

Application

Kovac′s Indole reagent is used for the detection of indole producing microorganisms in food and beverages, and environmental samples. This is suitable for the identification of E. coli.

Storage and Stability

No metal containers. Keep the container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition

Analysis Note

Indole formation (Escherichia coli ATCC 25922 (WDCM 00013)): +
Indole formation (Proteus vulgaris ATCC 8427): +
Indole formation (Enterobacter aerogenes ATCC 13048): -
Tested in DEV-Tryptophan broth art. no. 1.10694. (after incubation 24 hrs.; 35 °C aerobic).

Other Notes

The reagent solution must be stored in the dark in the refrigerator, otherwise it may turn brown and cannot be used.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

96.8 °F

Flash Point(C)

36 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

EN ISO 6579-1 standard specifies detection of Salmonella spp. in human consumption, animal feed, and environmental samples.

EN ISO 6579-1 standard specifies detection of Salmonella spp. in human consumption, animal feed, and environmental samples.

EN ISO 6579-1 standard specifies detection of Salmonella spp. in human consumption, animal feed, and environmental samples.

EN ISO 6579-1 standard specifies detection of Salmonella spp. in human consumption, animal feed, and environmental samples.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service