1.08046
Phosphate Test Refill pack
for use with MCOLORTEST®
Synonym(s):
Phosphate Visual Test Kit Refills
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116116
NACRES:
NA.24
Recommended Products
feature
refill
Quality Level
specific analyte(s)
phosphate
compatibility
for use with MCOLORTEST®
detection method
colorimetric
storage temp.
15-25°C
General description
This refill pack contains all the reagents for the MQuant® test kit 1.11138 for the colorimetric evaluation of phosphate. When the reagents of the MQuant® test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MQuant® test kits, making it a particularly economical and ecological system.
Method
In sulfuric solution orthophosphate ions (PO03-) react with molybdate ions to form molybdophosphoric acid. This is reduced to phosphomolybdenum blue (PMB). The phosphate concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color zones of a color-matching vessel.
Method
In sulfuric solution orthophosphate ions (PO03-) react with molybdate ions to form molybdophosphoric acid. This is reduced to phosphomolybdenum blue (PMB). The phosphate concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color zones of a color-matching vessel.
Legal Information
MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service