Skip to Content
Merck
All Photos(4)

Documents

1.07023

Supelco

Hexane

≥98.5% (Σ hexane isomers + methylcyclopentane ), EMPARTA® ACS reagent

Synonym(s):

Hexane, n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-777-6
NACRES:
NB.21

product name

n-Hexane, for analysis EMPARTA® ACS

grade

ACS reagent

Quality Level

vapor density

~3 (vs air)

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

product line

EMPARTA®

Assay

≥98.5% (Σ hexane isomers + methylcyclopentane )

form

liquid

autoignition temp.

453 °F

potency

25000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

7.7 %

impurities

≤0.0003 meq/g Water-soluble titrable acid
≤0.005% Sulfur compounds (as S)
≤0.02% Water

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.375 (lit.)

kinematic viscosity

0.50 cSt(20 °C)

bp

69 °C (lit.)

mp

−95 °C (lit.)

transition temp

flash point -22 °C

density

0.659 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMD Millipore offers EMPARTA grade solvents for routine tasks in analytical laboratories. EMPARTA grade solvents offer fewer test parameters than EMSURE, yet are still of high quality. EMPARTA solvents meet ACS requirements, making them ideal for a wide range of analytical applications.

Application


  • Chiral-induced synthesis of chiral covalent organic frameworks core-shell microspheres for HPLC enantioseparation: This research demonstrates the use of n-Hexane in the synthesis of advanced materials for chromatographic applications, showcasing its role in enhancing the efficiency of enantioseparation processes in pharmaceutical and chemical research (Liu et al., 2024).

  • Deep eutectic solvents: Preparation, properties, and food applications: The article reviews the applications of n-Hexane in the food industry, particularly its use in combination with deep eutectic solvents to enhance food processing techniques, demonstrating its versatility as an industrial solvent (Negi et al., 2024).

  • An efficient method for the preparative isolation and purification of alkaloids from Gelsemium by using high speed counter-current chromatography and preparative HPLC: This publication showcases the role of n-Hexane in the extraction and purification of bioactive compounds, underlining its critical importance in the pharmacological and biochemical sectors (Liang et al., 2024).

  • Application of ionic liquid extractant in enhanced separation of 2-propanol-n-hexane azeotrope system: Highlights the use of n-Hexane in advanced separation processes, specifically in azeotropic distillation, which is crucial for the chemical and environmental industries, demonstrating its utility in complex separation scenarios (Li et al., 2024).

Analysis Note

Purity Σ hexane isomers + methylcyclopentane (GC): ≥ 98.5 %
Identity (IR): conforms
Color: ≤ 10 Hazen
Water-soluble titrable acid: ≤ 0.0003 meq/g
Thiophene: conforms
Sulfur compounds (as S): ≤ 0.005 %
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.02 %

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service