Skip to Content
Merck
All Photos(1)

Documents

1.06682

Supelco

Sodium ammonium hydrogen phosphate tetrahydrate

for analysis EMSURE®

Synonym(s):

Sodium ammonium hydrogen phosphate tetrahydrate, Ammonium sodium hydrogen phosphate, Microcosmic salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H5NNaO4P · 4H2O
CAS Number:
Molecular Weight:
209.07
MDL number:
UNSPSC Code:
12352302
EC Index Number:
235-860-8
NACRES:
NA.21

Quality Level

product line

EMSURE®

Assay

≥99.0% (acidimetric)

form

solid

pH

8.3 (20 °C, 50 g/L in H2O)

mp

79 °C

solubility

167 g/L

density

0.85 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

As: ≤0.00005%
Fe: ≤0.0005%
K: ≤0.001%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

InChI

1S/H3N.Na.H3O4P.4H2O/c;;1-5(2,3)4;;;;/h1H3;;(H3,1,2,3,4);4*1H2/q;+1;;;;;/p-1

InChI key

IQTQISLCLWJRPM-UHFFFAOYSA-M

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Chloride (Cl): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.0005 %
As (Arsenic): ≤ 0.00005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service