Skip to Content
Merck
All Photos(2)

Key Documents

1.06575

Supelco

di-Sodium hydrogen phosphate heptahydrate

for analysis EMSURE® ACS

Synonym(s):

Sodium phosphate dibasic heptahydrate, Disodium hydrogen phosphate heptahydrate, Disodium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4 · 7H2O
CAS Number:
Molecular Weight:
268.07
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-448-7
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
154 g/L
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

4.9 (vs air)

product line

EMSURE®

Assay

98.0-102.0% (acidimetric)

form

solid

potency

12930 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter

pH

8.7-9.3 (25 °C, 50 g/L in H2O)

mp

48 °C (Elimination of water of crystallisation)

solubility

154 g/L

density

1.68 g/mL at 25 °C (lit.)

bulk density

800 kg/m3

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
heavy metals: ≤0.001%

storage temp.

2-30°C

SMILES string

O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.7H2O/c;;1-5(2,3)4;;;;;;;/h;;(H3,1,2,3,4);7*1H2/q2*+1;;;;;;;;/p-2

InChI key

PYLIXCKOHOHGKQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Linkage

Replaces: SX0715; SX0715-3; SX0715-20

Analysis Note

Assay (acidimetric): 98.0 - 102.0 %
Identity: passes test
Insoluble matter: ≤ 0.005 %
pH-value (5 %; water, 25 °C): 8.7 - 9.3
Chloride (Cl): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.005 %
Heavy metals (ACS): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Corresponds to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

Validate urine sample cleanup with HLB SPE for opioid analysis via LC-MS/MS using the Supel Swift HLB 96-well plate.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service