Skip to Content
Merck
All Photos(1)

Key Documents

1.06563

Supelco

Sodium peroxide

granular for analysis ACS,ISO

Synonym(s):

Sodium peroxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Na2O2
CAS Number:
Molecular Weight:
77.98
MDL number:
UNSPSC Code:
12352304
EC Index Number:
215-209-4
NACRES:
NB.21
grade:
ACS reagent
form:
solid
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. ISO

form

solid

pH

12.8 (20 °C, 100 g/L in H2O)

mp

460 °C

density

2.8 g/cm3 at 20 °C

bulk density

1138 kg/m3

storage temp.

15-25°C

SMILES string

[Na+].[Na+].[O-][O-]

InChI

1S/2Na.O2/c;;1-2/q2*+1;-2

InChI key

PFUVRDFDKPNGAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

Replaces: SX0700-6

Analysis Note

Assay (manganometric): ≥ 95.0 %
Identity: passes test
Chloride (Cl): ≤ 0.002 %
Phosphate (PO₄): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.001 %
Total nitrogen (N): ≤ 0.003 %
Heavy metals (as Pb): ≤ 0.002 %
Al (Aluminium): ≤ 0.001 %
Fe (Iron): ≤ 0.002 %

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 1 - Skin Corr. 1A

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service