Skip to Content
Merck
All Photos(4)

Key Documents

1.06012

Supelco

Methanol

≥99.9% (GC), anhydrous, reagent grade, for extraction and analytical purposes

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-659-6
NACRES:
NB.21
grade:
anhydrous
for extraction
Assay:
≥99.9% (GC)
application(s):
pharmaceutical
bp:
64.7 °C (lit.)
vapor pressure:
128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)
Pricing and availability is not currently available.

Product Name

Methanol, anhydrous for analysis (max. 0.003% H₂O)

grade

anhydrous
for extraction

Quality Level

vapor density

1.11 (vs air)

vapor pressure

128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

description

for analytical purposes

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

725 °F

potency

5628 mg/kg LD50, oral (Rat)
17100 mg/kg LD50, skin (Rabbit)

expl. lim.

36 %

impurities

≤0.0002 meq/g Alkalinity
≤0.0005 meq/g Acidity
≤0.003% Water
≤0.05% Acetone (GC)
≤0.1% Ethanol (GC)

evapn. residue

≤10 mg/L

color

APHA: ≤10

refractive index

n20/D 1.329 (lit.)

kinematic viscosity

0.54-0.59 cSt(20 °C)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

2-30°C

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application



  • Conductometric investigation of nano MnSO(4) association parameters and complex formation with glycylglycine in aqueous and methanol-water mixtures: Thermodynamic parameters and potential applications.: This study explores the conductometric properties of nano manganese sulfate in both aqueous and methanol-water mixtures, emphasizing thermodynamic parameters and potential analytical applications (Rashad et al., 2024).


  • Electrocatalysis Boosts the Methanol Thermocatalytic Dehydrogenation for High-Purity H(2) and CO Production.: This research highlights the role of methanol in thermocatalytic dehydrogenation processes, crucial for generating high-purity hydrogen and carbon monoxide, showcasing its significance in chemical synthesis and analytical chemistry (Wu et al., 2024).


Linkage

Discover EMSURE - to be more than sure!

Analysis Note

Evaporation Number: 1.9

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. HI IS this bottle sealed with a rubber seal/septa to keep it dry? So that I have to use a needle to get my methanol?

    1 answer
    1. This bottle does not have a septum cap, for which you would need a needle to access. If you are looking for a methanol product which does have a septum cap, please see products 322415 and 322415.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service