Skip to Content
Merck
All Photos(1)

Key Documents

1.05827

Supelco

Magnesium hydroxide carbonate

for analysis EMSURE®

Synonym(s):

Magnesium hydroxide carbonate, Magnesium carbonate basic

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352302
EC Index Number:
235-192-7

Quality Level

product line

EMSURE®

form

solid

concentration

≥24% (Mg, complexometric)

impurities

≤0.001% Total nitrogen (N)
≤0.005% Substances insoluble in hydrochloric acid
≤0.5% Substances soluble in water

pH

10.5 (20 °C, 50 g/L in H2O, suspension)

mp

600 °C (decomposition)

solubility

water: insoluble at 20 °C

density

2.16 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.003%

cation traces

Ba + Sr (as Ba): ≤0.001%
Ca: ≤0.01%
Cu: ≤0.0005%
Fe: ≤0.001%
K: ≤0.001%
Na: ≤0.2%
Pb: ≤0.001%
Zn: ≤0.0005%
heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

InChI

1S/CH2O3.Mg.H2O/c2-1(3)4;;/h(H2,2,3,4);;1H2/q;+2;/p-3

InChI key

OUHCLAKJJGMPSW-UHFFFAOYSA-K

Analysis Note

Assay (komplexometric, Mg): ≥ 24.0 %
Substances soluble in water: ≤ 0.5 %
Substances insoluble in hydrochloric acid: ≤ 0.005 %
Chloride (Cl): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.003 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.001 %
Barium and Strontium (as Ba): ≤ 0.001 %
Ca (Calcium): ≤ 0.01 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.001 %
Na (Sodium): ≤ 0.2 %
Pb (Lead): ≤ 0.001 %
Zn (Zinc): ≤ 0.0005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service